Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005108.3 | NP_005099.2 | 11 - 12 | 998 | 66 |
XM_011534325.2 | XP_011532627.1 | 11 - 12 | 979 | 66 | |
XM_011534327.1 | XP_011532629.1 | 11 - 12 | 979 | 66 | |
XM_011534328.2 | XP_011532630.1 | 11 - 12 | 979 | 66 | |
XM_011534329.1 | XP_011532631.1 | 11 - 12 | 979 | 66 | |
XM_011534330.2 | XP_011532632.1 | 11 - 12 | 979 | 66 | |
XM_017007595.1 | XP_016863084.1 | 9 - 10 | 747 | 66 | |
XM_017007596.1 | XP_016863085.1 | 11 - 12 | 1024 | 66 | |
XM_017007598.1 | XP_016863087.1 | 11 - 12 | 979 | 66 | |
XM_017007599.1 | XP_016863088.1 | 11 - 12 | 979 | 66 | |
GenBank mRNA | AB015046.1 | - | 11 - 12 | 1018 | 66 |
AK001205.1 | - | 4 - 5 | 269 | 66 | |
AK025728.1 | - | 11 - 12 | 950 | 66 | |
AK293325.1 | - | 9 - 10 | 752 | 66 | |
AK314386.1 | - | 11 - 12 | 946 | 66 | |
BC137076.1 | - | 11 - 12 | 930 | 66 | |
BC137080.1 | - | 11 - 12 | 930 | 66 |