Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207354.2 | NP_997237.2 | 14 - 15 | 1778 | 60 |
NR_030767.1 | 15 - 16 | 1861 | 60 | ||
XM_011544985.1 | XP_011543287.1 | 15 - 16 | 1868 | 60 | |
XM_011544986.1 | XP_011543288.1 | 16 - 17 | 2008 | 60 | |
XM_011544987.2 | XP_011543289.1 | 15 - 16 | 1842 | 60 | |
XM_011544988.2 | XP_011543290.1 | 10 - 11 | 1198 | 60 | |
GenBank mRNA | AK027313.1 | - | 7 - 8 | 1719 | 60 |
AK057681.1 | - | 7 - 8 | 2392 | 60 | |
AK091614.1 | - | 2 - 3 | 2811 | 60 | |
AK096277.1 | - | 14 - 15 | 1691 | 60 | |
AK124721.1 | - | 15 - 16 | 1858 | 60 | |
AK126438.1 | - | 13 - 14 | 1775 | 60 | |
BC044239.1 | - | 4 - 5 | 749 | 60 | |
BC110419.1 | - | 14 - 15 | 1751 | 60 | |
BC121024.1 | - | 14 - 15 | 1652 | 60 | |
BC121025.2 | - | 14 - 15 | 1652 | 60 |