Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41612679-41613176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT16
Entrez Gene ID
3868
Gene Name
keratin 16
Gene Aliases
CK16, FNEPPK, K16, K1CP, KRT16A, NEPPK, PC1
UniGene
Hs.655160
Gene Chromosome Location
Chr.17: 41609778-41612827 on Build GRCh38
Transcripts
NM_005557.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
321305 c.8C>A p.T3N Chr.17: 41612681-41612681 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs978333073 Chr.17: 41612843-41612843 on Build GRCh38
rs113878014 Chr.17: 41612767-41612767 on Build GRCh38
rs561311752 Chr.17: 41613113-41613113 on Build GRCh38
rs779888168 Chr.17: 41612706-41612706 on Build GRCh38
rs945437053 Chr.17: 41612826-41612826 on Build GRCh38
rs989969400 Chr.17: 41612982-41612982 on Build GRCh38
rs371495992 Chr.17: 41612761-41612761 on Build GRCh38
rs768296966 Chr.17: 41612712-41612712 on Build GRCh38
rs978112234 Chr.17: 41612996-41612996 on Build GRCh38
rs930864717 Chr.17: 41613081-41613081 on Build GRCh38
rs568480940 Chr.17: 41612870-41612870 on Build GRCh38
rs368214153 Chr.17: 41612965-41612965 on Build GRCh38
rs975029386 Chr.17: 41613048-41613048 on Build GRCh38
rs146866851 Chr.17: 41612962-41612962 on Build GRCh38
rs771019093 Chr.17: 41613052-41613052 on Build GRCh38
rs981938633 Chr.17: 41612727-41612728 on Build GRCh38
rs909660785 Chr.17: 41613121-41613121 on Build GRCh38
rs750850030 Chr.17: 41612700-41612700 on Build GRCh38
rs915187238 Chr.17: 41612771-41612771 on Build GRCh38
rs112427050 Chr.17: 41613080-41613080 on Build GRCh38
rs552817479 Chr.17: 41612856-41612856 on Build GRCh38
rs6503645 Chr.17: 41612949-41612949 on Build GRCh38
rs755692257 Chr.17: 41612703-41612703 on Build GRCh38
rs932357478 Chr.17: 41612890-41612890 on Build GRCh38
rs1048610893 Chr.17: 41613094-41613094 on Build GRCh38
rs763207283 Chr.17: 41612718-41612718 on Build GRCh38
rs535145211 Chr.17: 41612862-41612862 on Build GRCh38
rs570940743 Chr.17: 41612857-41612857 on Build GRCh38
rs188703511 Chr.17: 41612865-41612865 on Build GRCh38
rs944910568 Chr.17: 41613044-41613044 on Build GRCh38
rs796977554 Chr.17: 41612961-41612961 on Build GRCh38
rs1003868383 Chr.17: 41612940-41612940 on Build GRCh38
rs558365294 Chr.17: 41613043-41613043 on Build GRCh38
rs112338618 Chr.17: 41612766-41612766 on Build GRCh38
rs748762166 Chr.17: 41612711-41612711 on Build GRCh38
rs1033749371 Chr.17: 41612759-41612759 on Build GRCh38
rs989588698 Chr.17: 41612769-41612769 on Build GRCh38
rs535489860 Chr.17: 41612871-41612871 on Build GRCh38
rs939669839 Chr.17: 41613147-41613147 on Build GRCh38
rs576844204 Chr.17: 41613053-41613053 on Build GRCh38
rs781240174 Chr.17: 41613156-41613156 on Build GRCh38
rs200187418 Chr.17: 41612752-41612752 on Build GRCh38
rs549024044 Chr.17: 41612989-41612989 on Build GRCh38
rs554396402 Chr.17: 41612924-41612924 on Build GRCh38
rs865833872 Chr.17: 41612788-41612788 on Build GRCh38
rs966690692 Chr.17: 41612983-41612983 on Build GRCh38
rs888106866 Chr.17: 41612934-41612935 on Build GRCh38
rs1003429508 Chr.17: 41612958-41612958 on Build GRCh38

View Full Product Details