Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256850.1 | NP_001243779.1 | 2 - 3 | 310 | 62 |
NM_001267550.2 | NP_001254479.2 | 2 - 3 | 310 | 62 | |
NM_003319.4 | NP_003310.4 | 2 - 3 | 310 | 62 | |
NM_133378.4 | NP_596869.4 | 2 - 3 | 310 | 62 | |
NM_133379.4 | NP_596870.2 | 2 - 3 | 310 | 62 | |
NM_133432.3 | NP_597676.3 | 2 - 3 | 310 | 62 | |
NM_133437.4 | NP_597681.4 | 2 - 3 | 310 | 62 | |
XM_017004819.1 | XP_016860308.1 | 2 - 3 | 310 | 62 | |
XM_017004820.1 | XP_016860309.1 | 2 - 3 | 310 | 62 | |
XM_017004821.1 | XP_016860310.1 | 2 - 3 | 310 | 62 | |
XM_017004822.1 | XP_016860311.1 | 2 - 3 | 310 | 62 | |
XM_017004823.1 | XP_016860312.1 | 2 - 3 | 310 | 62 | |
GenBank mRNA | AK056602.1 | - | 2 - 3 | 310 | 62 |
BC013396.2 | - | 2 - 3 | 295 | 62 | |
BC058824.1 | - | 2 - 3 | 313 | 62 | |
BC070170.1 | - | 2 - 3 | 310 | 62 | |
X90568.1 | - | 2 - 3 | 217 | 62 |