Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_002196.2 | 1 - 2 | 1359 | 94 | |
NR_131223.1 | 1 - 2 | 1359 | 94 | ||
GenBank mRNA | AK092486.1 | - | 2 - 3 | 810 | 94 |
AK123560.1 | - | 1 - 2 | 85 | 94 | |
BC013067.1 | - | 1 - 2 | 201 | 94 | |
BC023213.1 | - | 1 - 2 | 671 | 94 | |
BC098439.1 | - | 1 - 2 | 719 | 94 | |
BC110657.1 | - | 1 - 2 | 958 | 94 | |
BE730693.1 | - | 1 - 2 | 237 | 94 |