Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286657.1 | NP_001273586.1 | 3 - 4 | 604 | 80 |
NM_001286661.1 | NP_001273590.1 | 2 - 3 | 304 | 80 | |
NM_152417.2 | NP_689630.1 | 4 - 5 | 743 | 80 | |
XM_005251150.1 | XP_005251207.1 | 4 - 5 | 732 | 80 | |
XM_011517454.1 | XP_011515756.1 | 4 - 5 | 756 | 80 | |
XM_011517455.2 | XP_011515757.1 | 4 - 5 | 732 | 80 | |
XM_017013040.1 | XP_016868529.1 | 3 - 4 | 316 | 80 | |
GenBank mRNA | AK056932.1 | - | 4 - 5 | 662 | 80 |
AL832935.1 | - | 2 - 3 | 213 | 80 | |
CN272366.1 | - | 2 - 3 | 231 | 80 |