Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170753.1 | NP_001164224.1 | 4 - 5 | 515 | 75 |
NM_145017.2 | NP_659454.2 | 4 - 5 | 515 | 75 | |
GenBank mRNA | AK057333.1 | - | 4 - 5 | 508 | 75 |
BC043246.1 | - | 4 - 5 | 513 | 75 | |
BC053995.1 | - | 3 - 4 | 412 | 75 | |
BI561157.1 | - | 4 - 5 | 512 | 75 | |
DB047394.1 | - | 4 - 5 | 515 | 75 | |
JF442033.1 | - | 4 - 5 | 488 | 75 |