Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9739628-9740133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR143
Entrez Gene ID
4935
Gene Name
G protein-coupled receptor 143
Gene Aliases
NYS6, OA1
UniGene
Hs.74124
Gene Chromosome Location
Chr.X: 9725413-9765965 on Build GRCh38
Transcripts
NM_000273.2 XM_005274541.3

SNP Detail

SNP ID Chromosome Location
rs1031183031 Chr.X: 9740096-9740096 on Build GRCh38
rs938800297 Chr.X: 9739862-9739862 on Build GRCh38
rs950502690 Chr.X: 9739829-9739829 on Build GRCh38
rs768071575 Chr.X: 9740064-9740064 on Build GRCh38
rs149886683 Chr.X: 9739760-9739760 on Build GRCh38
rs756686806 Chr.X: 9739721-9739722 on Build GRCh38
rs776710609 Chr.X: 9739671-9739671 on Build GRCh38
rs768774548 Chr.X: 9739665-9739665 on Build GRCh38
rs764137296 Chr.X: 9739967-9739967 on Build GRCh38
rs766347844 Chr.X: 9739680-9739680 on Build GRCh38
rs144886665 Chr.X: 9739925-9739925 on Build GRCh38
rs940698631 Chr.X: 9739926-9739926 on Build GRCh38
rs966513337 Chr.X: 9739934-9739934 on Build GRCh38
rs968984180 Chr.X: 9739773-9739773 on Build GRCh38
rs759315924 Chr.X: 9739655-9739655 on Build GRCh38
rs750910171 Chr.X: 9740111-9740111 on Build GRCh38
rs763168997 Chr.X: 9739672-9739672 on Build GRCh38
rs761416202 Chr.X: 9739968-9739968 on Build GRCh38
rs190882588 Chr.X: 9740029-9740029 on Build GRCh38
rs771928397 Chr.X: 9739654-9739654 on Build GRCh38
rs764944881 Chr.X: 9740053-9740053 on Build GRCh38
rs182953696 Chr.X: 9739904-9739904 on Build GRCh38
rs753117759 Chr.X: 9739756-9739756 on Build GRCh38
rs983554306 Chr.X: 9739853-9739853 on Build GRCh38
rs1050296384 Chr.X: 9740027-9740027 on Build GRCh38
rs767931116 Chr.X: 9739753-9739753 on Build GRCh38
rs924123732 Chr.X: 9740079-9740079 on Build GRCh38
rs759639368 Chr.X: 9739699-9739699 on Build GRCh38
rs185924882 Chr.X: 9739971-9739971 on Build GRCh38
rs927480865 Chr.X: 9739832-9739832 on Build GRCh38
rs946991296 Chr.X: 9740061-9740061 on Build GRCh38
rs62635047 Chr.X: 9739673-9739674 on Build GRCh38
rs774656143 Chr.X: 9739692-9739692 on Build GRCh38
rs1030711663 Chr.X: 9739662-9739662 on Build GRCh38
rs769479021 Chr.X: 9739749-9739749 on Build GRCh38
rs866294644 Chr.X: 9739698-9739698 on Build GRCh38
rs746959492 Chr.X: 9739660-9739660 on Build GRCh38
rs980350774 Chr.X: 9739796-9739796 on Build GRCh38

View Full Product Details