Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303512.1 | NP_001290441.1 | 1 - 2 | 329 | 67 |
NM_001303513.1 | NP_001290442.1 | 1 - 2 | 329 | 67 | |
NM_032512.3 | NP_115901.2 | 1 - 2 | 329 | 67 | |
GenBank mRNA | AB103478.1 | - | 1 - 2 | 310 | 67 |
AK091444.1 | - | 1 - 2 | 55 | 67 | |
AK127016.1 | - | 1 - 2 | 329 | 67 | |
AK309212.1 | - | 1 - 2 | 134 | 67 | |
BC117277.1 | - | 1 - 2 | 121 | 67 | |
BC143545.1 | - | 1 - 2 | 120 | 67 |