Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080571.1 | NP_542138.1 | 2 - 3 | 241 | 69 |
NM_153269.2 | NP_695001.2 | 2 - 3 | 237 | 69 | |
GenBank mRNA | AK091924.1 | - | 1 - 2 | 166 | 69 |
AK097394.1 | - | 2 - 3 | 208 | 69 | |
BC051762.1 | - | 1 - 2 | 186 | 69 | |
BC134417.1 | - | 2 - 3 | 195 | 69 | |
BC137450.1 | - | 2 - 3 | 204 | 69 | |
BC144649.1 | - | 2 - 3 | 208 | 69 | |
BX640715.1 | - | 1 - 2 | 181 | 69 | |
DB226405.1 | - | 2 - 3 | 237 | 69 |