Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040666.1 | NP_001035756.1 | 6 - 7 | 1520 | 66 |
NM_001244945.1 | NP_001231874.1 | 5 - 6 | 1726 | 66 | |
XM_017011956.1 | XP_016867445.1 | 6 - 7 | 1837 | 66 | |
XM_017011957.1 | XP_016867446.1 | 6 - 7 | 1639 | 66 | |
GenBank mRNA | AF526382.1 | - | 6 - 7 | 1520 | 66 |
AK092666.1 | - | 5 - 6 | 1668 | 66 | |
AY008443.1 | - | 6 - 7 | 1520 | 66 | |
AY358267.1 | - | 6 - 7 | 1520 | 66 |