Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132894588-132895093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSC1
Entrez Gene ID
7248
Gene Name
tuberous sclerosis 1
Gene Aliases
LAM, TSC
UniGene
Hs.370854
Gene Chromosome Location
Chr.9: 132891348-132945269 on Build GRCh38
Transcripts
NM_000368.4 NM_001162426.1 NM_001162427.1 XM_005272211.1 XM_006717271.1 XM_011518979.2 XM_017015096.1 XM_017015097.1 XM_017015098.1 XM_017015099.1 XM_017015100.1 XM_017015101.1

SNP Detail

SNP ID Chromosome Location
rs896054185 Chr.9: 132894939-132894939 on Build GRCh38
rs1040029736 Chr.9: 132894721-132894721 on Build GRCh38
rs185039865 Chr.9: 132894752-132894752 on Build GRCh38
rs1004755147 Chr.9: 132894918-132894918 on Build GRCh38
rs774716260 Chr.9: 132894911-132894911 on Build GRCh38
rs536391268 Chr.9: 132894745-132894745 on Build GRCh38
rs739442 Chr.9: 132894747-132894747 on Build GRCh38
rs536648576 Chr.9: 132894719-132894720 on Build GRCh38
rs925791599 Chr.9: 132894815-132894815 on Build GRCh38
rs397514841 Chr.9: 132894903-132894903 on Build GRCh38
rs886063607 Chr.9: 132894716-132894716 on Build GRCh38
rs886063606 Chr.9: 132894693-132894694 on Build GRCh38
rs756774572 Chr.9: 132894617-132894617 on Build GRCh38
rs72759433 Chr.9: 132894767-132894767 on Build GRCh38
rs552190342 Chr.9: 132894727-132894727 on Build GRCh38
rs2809244 Chr.9: 132894960-132894960 on Build GRCh38
rs979977228 Chr.9: 132894794-132894794 on Build GRCh38
rs932719047 Chr.9: 132894901-132894901 on Build GRCh38
rs983652318 Chr.9: 132894837-132894837 on Build GRCh38
rs540849136 Chr.9: 132894991-132894991 on Build GRCh38
rs944357809 Chr.9: 132894716-132894716 on Build GRCh38
rs79277527 Chr.9: 132894748-132894748 on Build GRCh38
rs896592064 Chr.9: 132894992-132894992 on Build GRCh38
rs773490178 Chr.9: 132894984-132894984 on Build GRCh38
rs575245339 Chr.9: 132894862-132894862 on Build GRCh38
rs907573896 Chr.9: 132894667-132894667 on Build GRCh38
rs739441 Chr.9: 132894728-132894728 on Build GRCh38
rs1013551475 Chr.9: 132894761-132894761 on Build GRCh38
rs748099884 Chr.9: 132894694-132894694 on Build GRCh38
rs58612431 Chr.9: 132894863-132894863 on Build GRCh38
rs1013888273 Chr.9: 132894754-132894754 on Build GRCh38
rs565416605 Chr.9: 132895041-132895041 on Build GRCh38
rs999882542 Chr.9: 132895003-132895003 on Build GRCh38
rs2809243 Chr.9: 132894913-132894913 on Build GRCh38
rs980032028 Chr.9: 132894683-132894683 on Build GRCh38
rs996918543 Chr.9: 132894722-132894722 on Build GRCh38
rs552958137 Chr.9: 132894766-132894766 on Build GRCh38
rs1001898393 Chr.9: 132895057-132895057 on Build GRCh38
rs905196379 Chr.9: 132895042-132895042 on Build GRCh38
rs768858045 Chr.9: 132894892-132894892 on Build GRCh38
rs886063608 Chr.9: 132894716-132894716 on Build GRCh38
rs1044312792 Chr.9: 132894736-132894736 on Build GRCh38
rs969974293 Chr.9: 132894783-132894783 on Build GRCh38
rs538239408 Chr.9: 132894773-132894773 on Build GRCh38
rs1033148358 Chr.9: 132895039-132895039 on Build GRCh38
rs760805774 Chr.9: 132894986-132894986 on Build GRCh38
rs556956686 Chr.9: 132894817-132894817 on Build GRCh38
rs1013608610 Chr.9: 132895020-132895020 on Build GRCh38
rs759483755 Chr.9: 132894614-132894614 on Build GRCh38
rs988676197 Chr.9: 132894823-132894823 on Build GRCh38
rs1027936609 Chr.9: 132894808-132894808 on Build GRCh38
rs749683153 Chr.9: 132894880-132894880 on Build GRCh38
rs932254275 Chr.9: 132894922-132894922 on Build GRCh38
rs886063605 Chr.9: 132894623-132894623 on Build GRCh38
rs911765877 Chr.9: 132894854-132894854 on Build GRCh38
rs397514790 Chr.9: 132894839-132894839 on Build GRCh38
rs944677799 Chr.9: 132894859-132894859 on Build GRCh38

View Full Product Details