Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132894968-132895465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSC1
Entrez Gene ID
7248
Gene Name
tuberous sclerosis 1
Gene Aliases
LAM, TSC
UniGene
Hs.370854
Gene Chromosome Location
Chr.9: 132891348-132945269 on Build GRCh38
Transcripts
NM_000368.4 NM_001162426.1 NM_001162427.1 XM_005272211.1 XM_006717271.1 XM_011518979.2 XM_017015096.1 XM_017015097.1 XM_017015098.1 XM_017015099.1 XM_017015100.1 XM_017015101.1

SNP Detail

SNP ID Chromosome Location
rs191614777 Chr.9: 132895429-132895429 on Build GRCh38
rs950263925 Chr.9: 132895259-132895259 on Build GRCh38
rs367801115 Chr.9: 132895083-132895083 on Build GRCh38
rs534651131 Chr.9: 132895405-132895405 on Build GRCh38
rs1048846161 Chr.9: 132895424-132895424 on Build GRCh38
rs758217339 Chr.9: 132895240-132895240 on Build GRCh38
rs189368676 Chr.9: 132895294-132895294 on Build GRCh38
rs1019174030 Chr.9: 132895387-132895387 on Build GRCh38
rs999829677 Chr.9: 132895308-132895308 on Build GRCh38
rs886063610 Chr.9: 132895235-132895235 on Build GRCh38
rs1043162638 Chr.9: 132895113-132895113 on Build GRCh38
rs149902841 Chr.9: 132895394-132895394 on Build GRCh38
rs886063612 Chr.9: 132895376-132895376 on Build GRCh38
rs975822024 Chr.9: 132895195-132895195 on Build GRCh38
rs540849136 Chr.9: 132894991-132894991 on Build GRCh38
rs575246018 Chr.9: 132895393-132895393 on Build GRCh38
rs896592064 Chr.9: 132894992-132894992 on Build GRCh38
rs995530018 Chr.9: 132895411-132895411 on Build GRCh38
rs952023511 Chr.9: 132895105-132895105 on Build GRCh38
rs979471364 Chr.9: 132895279-132895279 on Build GRCh38
rs886063613 Chr.9: 132895411-132895411 on Build GRCh38
rs1052638228 Chr.9: 132895350-132895350 on Build GRCh38
rs999882542 Chr.9: 132895003-132895003 on Build GRCh38
rs565416605 Chr.9: 132895041-132895041 on Build GRCh38
rs565743323 Chr.9: 132895293-132895293 on Build GRCh38
rs139119563 Chr.9: 132895108-132895108 on Build GRCh38
rs984093580 Chr.9: 132895145-132895145 on Build GRCh38
rs117425923 Chr.9: 132895322-132895322 on Build GRCh38
rs752538602 Chr.9: 132895151-132895151 on Build GRCh38
rs905196379 Chr.9: 132895042-132895042 on Build GRCh38
rs1001898393 Chr.9: 132895057-132895057 on Build GRCh38
rs965813218 Chr.9: 132895160-132895160 on Build GRCh38
rs368824230 Chr.9: 132895312-132895312 on Build GRCh38
rs530042727 Chr.9: 132895185-132895185 on Build GRCh38
rs1033148358 Chr.9: 132895039-132895039 on Build GRCh38
rs1028832068 Chr.9: 132895416-132895416 on Build GRCh38
rs1013608610 Chr.9: 132895020-132895020 on Build GRCh38
rs1044646480 Chr.9: 132895274-132895274 on Build GRCh38
rs974654245 Chr.9: 132895078-132895078 on Build GRCh38
rs886063611 Chr.9: 132895315-132895315 on Build GRCh38
rs562950766 Chr.9: 132895183-132895183 on Build GRCh38
rs886063614 Chr.9: 132895418-132895418 on Build GRCh38
rs765244801 Chr.9: 132895075-132895075 on Build GRCh38
rs953026524 Chr.9: 132895269-132895269 on Build GRCh38
rs1032684508 Chr.9: 132895320-132895320 on Build GRCh38
rs1037599510 Chr.9: 132895223-132895223 on Build GRCh38
rs544273113 Chr.9: 132895129-132895129 on Build GRCh38
rs921636386 Chr.9: 132895247-132895247 on Build GRCh38
rs896015087 Chr.9: 132895258-132895258 on Build GRCh38
rs918075245 Chr.9: 132895384-132895384 on Build GRCh38
rs886063609 Chr.9: 132895146-132895146 on Build GRCh38

View Full Product Details