Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152455.3 | NP_689668.3 | 2 - 3 | 670 | 66 |
XM_006720401.3 | XP_006720464.1 | 3 - 4 | 1430 | 66 | |
XM_011521266.2 | XP_011519568.1 | 3 - 4 | 1110 | 66 | |
XM_017021949.1 | XP_016877438.1 | 2 - 3 | 1642 | 66 | |
GenBank mRNA | AF525399.1 | - | 2 - 3 | 520 | 66 |
AK093186.1 | - | 1 - 2 | 216 | 66 | |
AK122788.1 | - | 2 - 3 | 658 | 66 | |
BC062551.1 | - | 2 - 3 | 286 | 66 |