Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152405.4 | NP_689618.4 | 2 - 3 | 1756 | 76 |
XM_005248430.2 | XP_005248487.1 | 2 - 3 | 1756 | 76 | |
XM_011543155.2 | XP_011541457.1 | 2 - 3 | 1756 | 76 | |
GenBank mRNA | AK095189.1 | - | 1 - 2 | 157 | 76 |
BC130624.1 | - | 1 - 2 | 182 | 76 | |
CN372786.1 | - | 2 - 3 | 396 | 76 | |
DW425980.1 | - | 1 - 2 | 140 | 76 |