Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032279.3 | NP_115655.2 | 12 - 13 | 1779 | 89 |
XM_011513232.2 | XP_011511534.1 | 12 - 13 | 1526 | 89 | |
XM_017007318.1 | XP_016862807.1 | 12 - 13 | 1415 | 89 | |
XM_017007319.1 | XP_016862808.1 | 12 - 13 | 1526 | 89 | |
GenBank mRNA | AK095277.1 | - | 12 - 13 | 1564 | 89 |
AK096510.1 | - | 3 - 4 | 375 | 89 | |
AL512736.1 | - | 4 - 5 | 598 | 89 | |
AY823162.1 | - | 12 - 13 | 1456 | 89 | |
BC101496.1 | - | 12 - 13 | 1550 | 89 |