Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032279.3 | NP_115655.2 | 20 - 21 | 2756 | 127 |
XM_011513232.2 | XP_011511534.1 | 20 - 21 | 2503 | 127 | |
XM_017007318.1 | XP_016862807.1 | 20 - 21 | 2392 | 127 | |
XM_017007319.1 | XP_016862808.1 | 20 - 21 | 2503 | 127 | |
GenBank mRNA | AK095277.1 | - | 20 - 21 | 2541 | 127 |
AK096510.1 | - | 11 - 12 | 1352 | 127 | |
AY823162.1 | - | 20 - 21 | 2433 | 127 | |
BC101496.1 | - | 20 - 21 | 2527 | 127 |