Assay Design



Gene Transcripts

Gene Symbol
SLC36A4
Entrez Gene ID
120103
Gene Name
solute carrier family 36 member 4
Gene Aliases
PAT4
Chromosome Location
Chr.11: 93144171 - 93197983 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286139.1 NP_001273068.1 5 - 6 637 68
NM_152313.3 NP_689526.2 5 - 6 600 68
XM_017017174.1 XP_016872663.1 5 - 6 604 68
XM_017017175.1 XP_016872664.1 4 - 5 517 68
XM_017017176.1 XP_016872665.1 4 - 5 469 68
XM_017017177.1 XP_016872666.1 4 - 5 373 68
GenBank mRNA AK096251.1 - 5 - 6 636 68
AY162216.1 - 5 - 6 554 68
BC047374.1 - 5 - 6 567 68

View Full Product Details