Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015172.3 | NP_055987.2 | 11 - 12 | 1515 | 92 |
XM_005245015.2 | XP_005245072.1 | 11 - 12 | 1552 | 92 | |
XM_005245016.4 | XP_005245073.1 | 11 - 12 | 1532 | 92 | |
XM_005245017.2 | XP_005245074.1 | 11 - 12 | 1552 | 92 | |
XM_005245018.2 | XP_005245075.1 | 11 - 12 | 1527 | 92 | |
XM_005245019.2 | XP_005245076.1 | 11 - 12 | 1552 | 92 | |
XM_005245020.2 | XP_005245077.1 | 11 - 12 | 1552 | 92 | |
XM_005245023.2 | XP_005245080.1 | 11 - 12 | 1552 | 92 | |
XM_005245024.2 | XP_005245081.1 | 11 - 12 | 1552 | 92 | |
XM_005245025.2 | XP_005245082.1 | 11 - 12 | 1552 | 92 | |
XM_005245026.2 | XP_005245083.1 | 11 - 12 | 1552 | 92 | |
XM_005245027.2 | XP_005245084.1 | 11 - 12 | 1552 | 92 | |
XM_005245028.2 | XP_005245085.1 | 11 - 12 | 1552 | 92 | |
XM_005245029.2 | XP_005245086.1 | 11 - 12 | 1552 | 92 | |
XM_006711238.3 | XP_006711301.1 | 11 - 12 | 1532 | 92 | |
XM_017000760.1 | XP_016856249.1 | 11 - 12 | 1517 | 92 | |
XM_017000761.1 | XP_016856250.1 | 11 - 12 | 1552 | 92 | |
XM_017000762.1 | XP_016856251.1 | 11 - 12 | 1552 | 92 | |
GenBank mRNA | AF253978.1 | - | 10 - 11 | 1320 | 92 |
AF488829.1 | - | 10 - 11 | 1249 | 92 | |
AK298887.1 | - | - | 546 | 92 | |
AL096857.1 | - | 10 - 11 | 1306 | 92 | |
BC001260.1 | - | 11 - 12 | 1486 | 92 | |
BC026108.1 | - | 11 - 12 | 1492 | 92 | |
BC045713.1 | - | 11 - 12 | 1487 | 92 | |
BC058930.1 | - | 11 - 12 | 1498 | 92 | |
BC066968.1 | - | 11 - 12 | 1536 | 92 |