Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286840.1 | NP_001273769.1 | 12 - 13 | 1348 | 108 |
NM_001286842.1 | NP_001273771.1 | 7 - 8 | 714 | 108 | |
NM_015651.2 | NP_056466.1 | 12 - 13 | 1477 | 108 | |
XM_005251906.2 | XP_005251963.1 | 12 - 13 | 1285 | 108 | |
XM_011518509.2 | XP_011516811.1 | 12 - 13 | 1328 | 108 | |
XM_011518511.2 | XP_011516813.1 | 12 - 13 | 1811 | 108 | |
XM_017014612.1 | XP_016870101.1 | 12 - 13 | 1317 | 108 | |
XM_017014613.1 | XP_016870102.1 | 12 - 13 | 1790 | 108 | |
GenBank mRNA | AK122744.1 | - | 12 - 13 | 1337 | 108 |
AK302996.1 | - | 7 - 8 | 684 | 108 | |
AK303433.1 | - | 10 - 11 | 1022 | 108 | |
AL117477.1 | - | 9 - 10 | 905 | 108 | |
BC125076.1 | - | 11 - 12 | 1249 | 108 | |
BC125077.1 | - | 11 - 12 | 1249 | 108 | |
BX640713.1 | - | 12 - 13 | 1349 | 108 |