Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009555.3 | NP_001009555.3 | 13 - 14 | 2543 | 103 |
NM_001128923.1 | NP_001122395.1 | 12 - 13 | 2462 | 103 | |
NM_001128924.1 | NP_001122396.1 | 5 - 6 | 1465 | 103 | |
NM_001243349.1 | NP_001230278.1 | 12 - 13 | 2383 | 103 | |
XM_005262767.2 | XP_005262824.1 | 14 - 15 | 2404 | 103 | |
XM_011531646.1 | XP_011529948.1 | 12 - 13 | 2190 | 103 | |
XM_011531647.1 | XP_011529949.1 | 11 - 12 | 2082 | 103 | |
XM_011531648.1 | XP_011529950.1 | 14 - 15 | 2517 | 103 | |
XM_011531649.1 | XP_011529951.1 | 10 - 11 | 2013 | 103 | |
XM_017007778.1 | XP_016863267.1 | 14 - 15 | 2508 | 103 | |
XM_017007779.1 | XP_016863268.1 | 10 - 11 | 2004 | 103 | |
XM_017007780.1 | XP_016863269.1 | 13 - 14 | 2439 | 103 | |
XM_017007781.1 | XP_016863270.1 | 13 - 14 | 2434 | 103 | |
XM_017007782.1 | XP_016863271.1 | 12 - 13 | 2365 | 103 | |
XM_017007783.1 | XP_016863272.1 | 13 - 14 | 2335 | 103 | |
GenBank mRNA | AK092528.1 | - | 5 - 6 | 695 | 103 |
AK128493.1 | - | 5 - 6 | 1314 | 103 | |
AK291025.1 | - | 5 - 6 | 631 | 103 | |
AK294476.1 | - | 12 - 13 | 2383 | 103 | |
AL133047.1 | - | 3 - 4 | 307 | 103 | |
BC010110.2 | - | 5 - 6 | 736 | 103 | |
BC085613.1 | - | 4 - 5 | 522 | 103 | |
BC108890.1 | - | 6 - 7 | 1573 | 103 | |
BC108891.1 | - | 6 - 7 | 1573 | 103 | |
BC108892.1 | - | 6 - 7 | 1573 | 103 | |
BC108893.1 | - | 5 - 6 | 1465 | 103 | |
BX647422.1 | - | 13 - 14 | 2455 | 103 |