Assay Design



Gene Transcripts

Gene Symbol
SYNGAP1
Entrez Gene ID
8831
Gene Name
synaptic Ras GTPase activating protein 1
Gene Aliases
MRD5, RASA1, RASA5, SYNGAP
Chromosome Location
Chr.6: 33420070 - 33453689 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006772.2 NP_006763.2 10 - 11 1872 119
GenBank mRNA AK307888.1 - 10 - 11 1687 119
AL713634.1 - 3 - 4 528 119

View Full Product Details