Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181644.4 | NP_857595.3 | 9 - 10 | 1577 | 117 |
XM_011509229.2 | XP_011507531.1 | 8 - 9 | 1316 | 117 | |
GenBank mRNA | AK057733.1 | - | 5 - 6 | 647 | 117 |
AK090831.1 | - | 3 - 4 | 304 | 117 | |
AK091896.1 | - | 9 - 10 | 1518 | 117 | |
AK127321.1 | - | 1 - 2 | 1213 | 117 | |
AK293816.1 | - | 8 - 9 | 1506 | 117 | |
AK295148.1 | - | 8 - 9 | 1164 | 117 | |
AK304109.1 | - | 8 - 9 | 1382 | 117 | |
AK304120.1 | - | 8 - 9 | 1243 | 117 | |
AL713676.1 | - | 7 - 8 | 883 | 117 | |
AY358107.1 | - | 7 - 8 | 964 | 117 | |
BC036549.2 | - | 9 - 10 | 1527 | 117 |