Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153610.4 | NP_705838.3 | 7 - 8 | 11326 | 105 |
XM_017009212.1 | XP_016864701.1 | 7 - 8 | 11194 | 105 | |
GenBank mRNA | AF533705.1 | - | 6 - 7 | 1920 | 105 |
AL831966.1 | - | 6 - 7 | 3459 | 105 | |
AL831968.2 | - | 2 - 3 | 253 | 105 | |
AL831986.1 | - | 6 - 7 | 1878 | 105 | |
AL832021.1 | - | 6 - 7 | 3700 | 105 | |
AL832368.1 | - | 6 - 7 | 2933 | 105 | |
AL834252.2 | - | 6 - 7 | 3043 | 105 | |
BC111529.2 | - | 6 - 7 | 2636 | 105 | |
BC111530.2 | - | 6 - 7 | 2636 | 105 | |
BX647628.1 | - | 6 - 7 | 3030 | 105 | |
BX647646.1 | - | 6 - 7 | 3030 | 105 |