Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153610.4 | NP_705838.3 | 9 - 10 | 11625 | 91 |
XM_017009212.1 | XP_016864701.1 | 9 - 10 | 11493 | 91 | |
GenBank mRNA | AF533705.1 | - | 8 - 9 | 2219 | 91 |
AL359338.1 | - | 1 - 2 | 375 | 91 | |
AL831966.1 | - | 8 - 9 | 3758 | 91 | |
AL831968.2 | - | 4 - 5 | 552 | 91 | |
AL831986.1 | - | 7 - 8 | 3281 | 91 | |
AL832021.1 | - | 8 - 9 | 3999 | 91 | |
AL834252.2 | - | 8 - 9 | 3342 | 91 | |
BC111529.2 | - | 8 - 9 | 2935 | 91 | |
BC111530.2 | - | 8 - 9 | 2935 | 91 | |
BX647628.1 | - | 8 - 9 | 3329 | 91 | |
BX647646.1 | - | 8 - 9 | 3329 | 91 |