Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173854.5 | NP_776253.3 | 10 - 11 | 2393 | 92 |
XM_005245069.2 | XP_005245126.1 | 10 - 11 | 2069 | 92 | |
GenBank mRNA | AJ514402.1 | - | 9 - 10 | 1480 | 92 |
AK093254.1 | - | 2 - 3 | 1414 | 92 | |
AK125075.1 | - | 9 - 10 | 1154 | 92 | |
AK128069.1 | - | 9 - 10 | 1646 | 92 | |
AK314732.1 | - | 9 - 10 | 1551 | 92 | |
AL831974.1 | - | 10 - 11 | 2229 | 92 | |
AL832001.1 | - | 10 - 11 | 2229 | 92 | |
AL832362.1 | - | 10 - 11 | 2229 | 92 | |
BC136303.1 | - | 9 - 10 | 1874 | 92 | |
BX641054.1 | - | 10 - 11 | 2232 | 92 |