Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100588.1 | NP_001094058.1 | 1 - 2 | 178 | 87 |
NM_018835.2 | NP_061323.2 | 1 - 2 | 178 | 87 | |
GenBank mRNA | AL833177.1 | - | 1 - 2 | 117 | 87 |
BC044642.1 | - | 1 - 2 | 110 | 87 | |
BU783286.1 | - | 1 - 2 | 184 | 87 |