Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100588.1 | NP_001094058.1 | 12 - 13 | 2523 | 67 |
NM_018835.2 | NP_061323.2 | 12 - 13 | 2523 | 67 | |
GenBank mRNA | AF255303.1 | - | 11 - 12 | 2883 | 67 |
AK000308.1 | - | 10 - 11 | 1957 | 67 | |
AK027042.1 | - | 3 - 4 | 760 | 67 | |
AK226010.1 | - | 10 - 11 | 1957 | 67 | |
AL833177.1 | - | 13 - 14 | 2380 | 67 | |
AL834431.1 | - | 6 - 7 | 1274 | 67 |