Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128608.1 | NP_001122080.1 | 10 - 11 | 1281 | 76 |
NM_001265611.1 | NP_001252540.1 | 9 - 10 | 1263 | 76 | |
NM_014994.2 | NP_055809.2 | 9 - 10 | 1263 | 76 | |
NR_049762.1 | 8 - 9 | 1171 | 76 | ||
XM_006720438.1 | XP_006720501.1 | 9 - 10 | 1227 | 76 | |
XM_011521383.1 | XP_011519685.1 | 10 - 11 | 1245 | 76 | |
XM_011521384.2 | XP_011519686.1 | 10 - 11 | 1245 | 76 | |
XM_017022017.1 | XP_016877506.1 | 9 - 10 | 1521 | 76 | |
GenBank mRNA | AB011168.2 | - | 10 - 11 | 1191 | 76 |
AK091616.1 | - | 7 - 8 | 1933 | 76 | |
AK302482.1 | - | 8 - 9 | 1112 | 76 | |
AL833267.1 | - | 9 - 10 | 1183 | 76 | |
BC113983.1 | - | 9 - 10 | 1155 | 76 | |
BC114493.1 | - | 10 - 11 | 1173 | 76 | |
BX640787.1 | - | 7 - 8 | 1210 | 76 | |
BX648786.1 | - | 9 - 10 | 1214 | 76 |