Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011649.2 | NP_001011649.1 | 29 - 30 | 4604 | 93 |
NM_001272039.1 | NP_001258968.1 | 26 - 27 | 3914 | 93 | |
NM_018249.5 | NP_060719.4 | 29 - 30 | 4604 | 93 | |
NR_073554.1 | 29 - 30 | 4681 | 93 | ||
NR_073555.1 | 29 - 30 | 4604 | 93 | ||
NR_073556.1 | 30 - 31 | 4811 | 93 | ||
NR_073557.1 | 29 - 30 | 4684 | 93 | ||
NR_073558.1 | 29 - 30 | 4681 | 93 | ||
XM_006717182.1 | XP_006717245.1 | 28 - 29 | 4497 | 93 | |
XM_006717185.1 | XP_006717248.1 | 26 - 27 | 3906 | 93 | |
XM_011518860.1 | XP_011517162.1 | 29 - 30 | 4590 | 93 | |
XM_017014921.1 | XP_016870410.1 | 28 - 29 | 4494 | 93 | |
XM_017014922.1 | XP_016870411.1 | 23 - 24 | 3822 | 93 | |
XM_017014923.1 | XP_016870412.1 | 26 - 27 | 3906 | 93 | |
XM_017014924.1 | XP_016870413.1 | 20 - 21 | 3578 | 93 | |
GenBank mRNA | AB046853.2 | - | 29 - 30 | 4573 | 93 |
AF448860.1 | - | 29 - 30 | 4554 | 93 | |
AK001729.1 | - | 7 - 8 | 1442 | 93 | |
AK025867.1 | - | 2 - 3 | 171 | 93 | |
AK027636.1 | - | 8 - 9 | 1568 | 93 | |
AK122913.1 | - | 29 - 30 | 4593 | 93 | |
AL133161.1 | - | 13 - 14 | 2594 | 93 | |
AL833481.1 | - | 29 - 30 | 4592 | 93 | |
BC004526.2 | - | 2 - 3 | 213 | 93 | |
BC136275.1 | - | 29 - 30 | 4436 | 93 | |
BC143762.1 | - | 29 - 30 | 4439 | 93 | |
BC146782.1 | - | 29 - 30 | 4573 | 93 | |
BX537421.1 | - | 29 - 30 | 4589 | 93 | |
BX537708.1 | - | 26 - 27 | 3903 | 93 | |
BX537759.1 | - | 29 - 30 | 4592 | 93 | |
BX640896.1 | - | 22 - 23 | 3720 | 93 | |
CR936622.1 | - | 22 - 23 | 3734 | 93 | |
CR936717.1 | - | 22 - 23 | 3734 | 93 | |
KP294527.1 | - | 29 - 30 | 4604 | 93 |