Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001265612.1 | NP_001252541.1 | 23 - 24 | 3520 | 77 |
NM_016284.4 | NP_057368.3 | 23 - 24 | 3535 | 77 | |
NM_206999.2 | NP_996882.1 | 23 - 24 | 3535 | 77 | |
NR_049763.1 | 23 - 24 | 3520 | 77 | ||
GenBank mRNA | AB023224.2 | - | 10 - 11 | 1580 | 77 |
AK056927.1 | - | 14 - 15 | 2070 | 77 | |
AL832769.1 | - | 23 - 24 | 3484 | 77 | |
AL833549.1 | - | 23 - 24 | 3495 | 77 | |
BC024317.1 | - | 6 - 7 | 934 | 77 | |
BC040523.1 | - | 23 - 24 | 3487 | 77 | |
BX537840.1 | - | 23 - 24 | 3466 | 77 | |
BX647815.1 | - | 23 - 24 | 3475 | 77 | |
EF553522.1 | - | 23 - 24 | 3469 | 77 |