Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014494.2 | NP_055309.2 | 17 - 18 | 4652 | 107 |
XM_005255254.3 | XP_005255311.1 | 16 - 17 | 4495 | 107 | |
XM_005255257.4 | XP_005255314.1 | 16 - 17 | 4619 | 107 | |
XM_017023143.1 | XP_016878632.1 | 16 - 17 | 4596 | 107 | |
XM_017023144.1 | XP_016878633.1 | 15 - 16 | 4563 | 107 | |
XM_017023145.1 | XP_016878634.1 | 16 - 17 | 4529 | 107 | |
XM_017023146.1 | XP_016878635.1 | 16 - 17 | 4661 | 107 | |
XM_017023147.1 | XP_016878636.1 | 16 - 17 | 4689 | 107 | |
XM_017023148.1 | XP_016878637.1 | 15 - 16 | 4416 | 107 | |
XM_017023149.1 | XP_016878638.1 | 14 - 15 | 4413 | 107 | |
XM_017023150.1 | XP_016878639.1 | 15 - 16 | 4378 | 107 | |
XM_017023151.1 | XP_016878640.1 | 14 - 15 | 4267 | 107 | |
XM_017023152.1 | XP_016878641.1 | 15 - 16 | 4168 | 107 | |
XM_017023153.1 | XP_016878642.1 | 16 - 17 | 4619 | 107 | |
XM_017023154.1 | XP_016878643.1 | 14 - 15 | 4510 | 107 | |
GenBank mRNA | AB040893.1 | - | 12 - 13 | 2852 | 107 |
AJ492221.1 | - | 16 - 17 | 4538 | 107 | |
AK025696.1 | - | 10 - 11 | 1209 | 107 | |
AL713750.1 | - | 4 - 5 | 1405 | 107 | |
AL833757.1 | - | 11 - 12 | 1813 | 107 | |
AY035864.1 | - | 14 - 15 | 4186 | 107 |