Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163034.1 | NP_001156506.1 | 10 - 11 | 2016 | 59 |
NM_020761.2 | NP_065812.1 | 10 - 11 | 2016 | 59 | |
GenBank mRNA | AB037724.1 | - | 6 - 7 | 698 | 59 |
AB082951.1 | - | 10 - 11 | 1211 | 59 | |
AK055912.1 | - | 10 - 11 | 1862 | 59 | |
AK295964.1 | - | 9 - 10 | 1939 | 59 | |
AK296321.1 | - | 10 - 11 | 2011 | 59 | |
AY090663.1 | - | 10 - 11 | 1573 | 59 | |
BC136652.1 | - | 10 - 11 | 1406 | 59 | |
BC136654.1 | - | 10 - 11 | 1406 | 59 |