Assay Design
Gene Transcripts
- Gene Symbol
- TAF4
- Entrez Gene ID
-
6874
- Gene Name
- TATA-box binding protein associated factor 4
- Gene Aliases
-
TAF2C, TAF2C1, TAF4A, TAFII130, TAFII135
- Chromosome Location
-
Chr.20: 61974798 - 62065810 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_003185.3 |
NP_003176.2 |
1 - 2 |
1361 |
87 |
GenBank mRNA |
U75308.1 |
- |
3 - 4 |
947 |
87 |
Y11354.1 |
- |
1 - 2 |
1355 |
87 |
View Full Product Details