Assay Design



Gene Transcripts

Gene Symbol
HUWE1
Entrez Gene ID
10075
Gene Name
HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
Gene Aliases
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1
Chromosome Location
Chr.X: 53532096 - 53686729 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031407.6 NP_113584.3 70 - 71 11320 79
XM_005261965.3 XP_005262022.1 69 - 70 11387 79
XM_011530747.1 XP_011529049.1 71 - 72 11563 79
XM_011530751.2 XP_011529053.1 70 - 71 11431 79
XM_017029191.1 XP_016884680.1 69 - 70 14315 79
XM_017029192.1 XP_016884681.1 69 - 70 14312 79
XM_017029193.1 XP_016884682.1 69 - 70 14294 79
XM_017029194.1 XP_016884683.1 69 - 70 14315 79
XM_017029195.1 XP_016884684.1 69 - 70 14267 79
XM_017029196.1 XP_016884685.1 69 - 70 14264 79
XM_017029197.1 XP_016884686.1 68 - 69 14216 79
XM_017029198.1 XP_016884687.1 70 - 71 11657 79
XM_017029199.1 XP_016884688.1 71 - 72 11584 79
XM_017029200.1 XP_016884689.1 70 - 71 11454 79
XM_017029201.1 XP_016884690.1 70 - 71 11471 79
XM_017029202.1 XP_016884691.1 70 - 71 11447 79
XM_017029203.1 XP_016884692.1 70 - 71 11453 79
XM_017029204.1 XP_016884693.1 68 - 69 14065 79
XM_017029205.1 XP_016884694.1 67 - 68 13943 79
XM_017029206.1 XP_016884695.1 66 - 67 13913 79
GenBank mRNA AB002310.3 - 39 - 40 7371 79
AB071605.1 - 43 - 44 8020 79
AF161390.1 - 16 - 17 3567 79
AY772009.1 - 68 - 69 10965 79
AY929612.1 - 67 - 68 10917 79
DQ097177.1 - 70 - 71 11319 79

View Full Product Details