Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139459.1 | NP_001132931.1 | 3 - 4 | 1095 | 69 |
NM_152609.2 | NP_689822.2 | 3 - 4 | 969 | 69 | |
XM_005273081.3 | XP_005273138.2 | 3 - 4 | 754 | 69 | |
XM_011544110.2 | XP_011542412.1 | 3 - 4 | 820 | 69 | |
XM_011544111.1 | XP_011542413.1 | 3 - 4 | 991 | 69 | |
XM_011544112.1 | XP_011542414.1 | 2 - 3 | 538 | 69 | |
XM_017000487.1 | XP_016855976.1 | 4 - 5 | 1009 | 69 | |
GenBank mRNA | AK074142.1 | - | 3 - 4 | 732 | 69 |
AL834246.2 | - | 3 - 4 | 803 | 69 | |
BC010228.2 | - | 3 - 4 | 971 | 69 | |
BC036200.1 | - | 3 - 4 | 862 | 69 | |
BX647629.1 | - | 3 - 4 | 754 | 69 |