Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014460.3 | NP_055275.1 | 3 - 4 | 593 | 61 |
GenBank mRNA | AB027011.1 | - | 2 - 3 | 316 | 61 |
AK313407.1 | - | 3 - 4 | 550 | 61 | |
AL834417.1 | - | 3 - 4 | 478 | 61 | |
BC067113.1 | - | 3 - 4 | 539 | 61 | |
CR456457.1 | - | 2 - 3 | 328 | 61 |