Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198578.3 | NP_940980.3 | 44 - 45 | 6699 | 90 |
XM_005268629.3 | XP_005268686.1 | 44 - 45 | 6815 | 90 | |
XM_011537877.2 | XP_011536179.1 | 44 - 45 | 6815 | 90 | |
XM_017018787.1 | XP_016874276.1 | 23 - 24 | 3643 | 90 | |
XM_017018788.1 | XP_016874277.1 | 19 - 20 | 2887 | 90 | |
GenBank mRNA | AK122640.1 | - | 11 - 12 | 1607 | 90 |
AK131537.1 | - | 19 - 20 | 3003 | 90 | |
AL832453.2 | - | 23 - 24 | 3872 | 90 | |
AL834529.1 | - | 1 - 2 | 197 | 90 | |
AY792511.1 | - | 44 - 45 | 6578 | 90 | |
BC117180.1 | - | 44 - 45 | 6636 | 90 |