Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015295.2 | NP_056110.2 | 2 - 3 | 447 | 112 |
XM_011525642.1 | XP_011523944.1 | 2 - 3 | 447 | 112 | |
XM_011525643.2 | XP_011523945.1 | 2 - 3 | 447 | 112 | |
XM_017025684.1 | XP_016881173.1 | 2 - 3 | 447 | 112 | |
GenBank mRNA | BC006008.1 | - | 2 - 3 | 447 | 112 |