Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015150.1 | NP_055965.1 | 3 - 4 | 614 | 108 |
XM_005264985.1 | XP_005265042.1 | 3 - 4 | 571 | 108 | |
XM_005264986.2 | XP_005265043.1 | 3 - 4 | 468 | 108 | |
XM_006713069.1 | XP_006713132.1 | 3 - 4 | 477 | 108 | |
XM_011533529.1 | XP_011531831.1 | 5 - 6 | 1011 | 108 | |
XM_011533530.1 | XP_011531832.1 | 3 - 4 | 639 | 108 | |
XM_011533531.1 | XP_011531833.1 | 5 - 6 | 1011 | 108 | |
XM_017006005.1 | XP_016861494.1 | 5 - 6 | 1011 | 108 | |
XM_017006006.1 | XP_016861495.1 | 5 - 6 | 1011 | 108 | |
XM_017006007.1 | XP_016861496.1 | 3 - 4 | 598 | 108 | |
GenBank mRNA | AK074446.1 | - | 3 - 4 | 419 | 108 |
AK301024.1 | - | 2 - 3 | 307 | 108 | |
AY237654.1 | - | 3 - 4 | 605 | 108 | |
BC032349.1 | - | 2 - 3 | 287 | 108 | |
BC051336.1 | - | 3 - 4 | 391 | 108 | |
BC069209.1 | - | 3 - 4 | 391 | 108 | |
BC098270.1 | - | 3 - 4 | 438 | 108 | |
BC100667.1 | - | 3 - 4 | 438 | 108 | |
BC100669.1 | - | 3 - 4 | 438 | 108 | |
BC121795.1 | - | 3 - 4 | 438 | 108 | |
D42043.1 | - | 3 - 4 | 542 | 108 |