Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014976.1 | NP_055791.1 | 8 - 9 | 1061 | 72 |
XM_005269647.3 | XP_005269704.1 | 8 - 9 | 1101 | 72 | |
XM_011539538.2 | XP_011537840.1 | 8 - 9 | 1049 | 72 | |
XM_011539539.2 | XP_011537841.1 | 8 - 9 | 1049 | 72 | |
XM_011539540.1 | XP_011537842.1 | 7 - 8 | 915 | 72 | |
GenBank mRNA | AK294971.1 | - | 8 - 9 | 1090 | 72 |
AK307418.1 | - | 8 - 9 | 1074 | 72 | |
BC049838.1 | - | 8 - 9 | 1062 | 72 | |
BC064486.1 | - | 8 - 9 | 1054 | 72 | |
BC080560.1 | - | 8 - 9 | 1054 | 72 | |
BC111040.1 | - | 8 - 9 | 1016 | 72 | |
D80007.1 | - | 8 - 9 | 1014 | 72 |