Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015155.2 | NP_055970.1 | 13 - 14 | 1792 | 73 |
XM_005252432.3 | XP_005252489.1 | 13 - 14 | 1801 | 73 | |
XM_005252435.3 | XP_005252492.1 | 13 - 14 | 1585 | 73 | |
XM_011519435.2 | XP_011517737.1 | 13 - 14 | 1636 | 73 | |
XM_017015987.1 | XP_016871476.1 | 12 - 13 | 3841 | 73 | |
XM_017015988.1 | XP_016871477.1 | 12 - 13 | 3839 | 73 | |
XM_017015989.1 | XP_016871478.1 | 12 - 13 | 3841 | 73 | |
XM_017015990.1 | XP_016871479.1 | 13 - 14 | 3360 | 73 | |
XM_017015992.1 | XP_016871481.1 | 8 - 9 | 1055 | 73 | |
XM_017015993.1 | XP_016871482.1 | 11 - 12 | 1441 | 73 | |
XM_017015994.1 | XP_016871483.1 | 12 - 13 | 1478 | 73 | |
GenBank mRNA | BC131630.1 | - | 12 - 13 | 1509 | 73 |
BC152443.1 | - | 12 - 13 | 1516 | 73 | |
D86971.2 | - | 12 - 13 | 1516 | 73 |