Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32578739-32579268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DRB1
Entrez Gene ID
3123
Gene Name
major histocompatibility complex, class II, DR beta 1
Gene Aliases
DRB1, DRw10, HLA-DR1B, HLA-DRB, SS1
UniGene
Hs.534322
Gene Chromosome Location
Chr.6: 32578769-32589836 on Build GRCh38
Transcripts
NM_001243965.1 NM_002124.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5766025 c.790T>C p.F264L Chr.6: 32579102-32579102 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778127920 Chr.6: 32578915-32578915 on Build GRCh38
rs3205684 Chr.6: 32578837-32578837 on Build GRCh38
rs796070117 Chr.6: 32578942-32578942 on Build GRCh38
rs113804375 Chr.6: 32578926-32578926 on Build GRCh38
rs1060190 Chr.6: 32578900-32578900 on Build GRCh38
rs1060129 Chr.6: 32578981-32578981 on Build GRCh38
rs768859416 Chr.6: 32579030-32579030 on Build GRCh38
rs777675672 Chr.6: 32579076-32579076 on Build GRCh38
rs34205910 Chr.6: 32578878-32578879 on Build GRCh38
rs771788701 Chr.6: 32579074-32579075 on Build GRCh38
rs118126743 Chr.6: 32578854-32578854 on Build GRCh38
rs754848562 Chr.6: 32579025-32579026 on Build GRCh38
rs1064707 Chr.6: 32578966-32578966 on Build GRCh38
rs202053852 Chr.6: 32579066-32579066 on Build GRCh38
rs201477797 Chr.6: 32579154-32579154 on Build GRCh38
rs1141883 Chr.6: 32578815-32578815 on Build GRCh38
rs116358897 Chr.6: 32579005-32579005 on Build GRCh38
rs35445101 Chr.6: 32579102-32579102 on Build GRCh38
rs868233377 Chr.6: 32579091-32579091 on Build GRCh38
rs1732 Chr.6: 32578934-32578934 on Build GRCh38
rs35263976 Chr.6: 32578865-32578865 on Build GRCh38
rs751986414 Chr.6: 32578995-32578995 on Build GRCh38
rs113734598 Chr.6: 32578984-32578984 on Build GRCh38
rs773293837 Chr.6: 32579010-32579010 on Build GRCh38
rs760009375 Chr.6: 32579128-32579128 on Build GRCh38
rs35324556 Chr.6: 32579042-32579042 on Build GRCh38
rs142078339 Chr.6: 32578949-32578949 on Build GRCh38
rs3200044 Chr.6: 32578962-32578962 on Build GRCh38
rs34542752 Chr.6: 32578875-32578875 on Build GRCh38
rs35569931 Chr.6: 32579198-32579198 on Build GRCh38
rs35000099 Chr.6: 32578924-32578924 on Build GRCh38
rs35521457 Chr.6: 32579033-32579033 on Build GRCh38
rs754083017 Chr.6: 32578903-32578903 on Build GRCh38
rs201099263 Chr.6: 32579078-32579078 on Build GRCh38
rs34900518 Chr.6: 32578850-32578850 on Build GRCh38
rs117417462 Chr.6: 32579120-32579120 on Build GRCh38
rs34656332 Chr.6: 32579218-32579218 on Build GRCh38
rs35388679 Chr.6: 32579116-32579116 on Build GRCh38
rs751231236 Chr.6: 32579152-32579152 on Build GRCh38
rs35716402 Chr.6: 32578930-32578930 on Build GRCh38
rs34215479 Chr.6: 32578851-32578851 on Build GRCh38
rs1064694 Chr.6: 32579061-32579061 on Build GRCh38
rs1064697 Chr.6: 32579051-32579051 on Build GRCh38
rs755803542 Chr.6: 32579021-32579024 on Build GRCh38
rs1060176 Chr.6: 32578927-32578927 on Build GRCh38
rs35165835 Chr.6: 32578961-32578962 on Build GRCh38
rs796894867 Chr.6: 32579076-32579078 on Build GRCh38
rs9269688 Chr.6: 32579023-32579023 on Build GRCh38
rs34923246 Chr.6: 32578847-32578847 on Build GRCh38
rs796968011 Chr.6: 32579061-32579062 on Build GRCh38
rs1059920 Chr.6: 32579072-32579072 on Build GRCh38
rs34958200 Chr.6: 32579157-32579157 on Build GRCh38
rs796682714 Chr.6: 32579158-32579158 on Build GRCh38
rs9269696 Chr.6: 32579173-32579173 on Build GRCh38
rs1064695 Chr.6: 32579056-32579056 on Build GRCh38
rs148808172 Chr.6: 32579186-32579186 on Build GRCh38
rs755300637 Chr.6: 32578904-32578904 on Build GRCh38
rs755057839 Chr.6: 32579112-32579112 on Build GRCh38
rs36217730 Chr.6: 32579041-32579041 on Build GRCh38
rs34828632 Chr.6: 32579153-32579153 on Build GRCh38
rs72847693 Chr.6: 32579201-32579201 on Build GRCh38
rs1060081 Chr.6: 32579004-32579004 on Build GRCh38
rs763256972 Chr.6: 32578903-32578904 on Build GRCh38
rs779655749 Chr.6: 32579019-32579019 on Build GRCh38
rs9269694 Chr.6: 32579141-32579141 on Build GRCh38
rs34266013 Chr.6: 32578813-32578813 on Build GRCh38
rs68069105 Chr.6: 32579078-32579078 on Build GRCh38
rs35340089 Chr.6: 32579151-32579151 on Build GRCh38
rs199703384 Chr.6: 32578877-32578878 on Build GRCh38
rs35662203 Chr.6: 32579122-32579122 on Build GRCh38
rs747615942 Chr.6: 32578916-32578916 on Build GRCh38
rs9269693 Chr.6: 32579089-32579089 on Build GRCh38
rs201375698 Chr.6: 32579065-32579065 on Build GRCh38
rs758801058 Chr.6: 32578911-32578911 on Build GRCh38
rs71864678 Chr.6: 32579022-32579022 on Build GRCh38
rs35297326 Chr.6: 32578802-32578802 on Build GRCh38
rs35236441 Chr.6: 32579034-32579034 on Build GRCh38
rs35467716 Chr.6: 32579144-32579144 on Build GRCh38
rs146292738 Chr.6: 32579044-32579044 on Build GRCh38
rs752464470 Chr.6: 32579043-32579043 on Build GRCh38
rs34839759 Chr.6: 32578780-32578780 on Build GRCh38
rs35195677 Chr.6: 32578905-32578905 on Build GRCh38
rs994092402 Chr.6: 32579076-32579076 on Build GRCh38
rs3180797 Chr.6: 32579110-32579110 on Build GRCh38
rs34834054 Chr.6: 32578768-32578768 on Build GRCh38
rs34007709 Chr.6: 32578991-32578991 on Build GRCh38
rs35528115 Chr.6: 32578764-32578764 on Build GRCh38
rs9281863 Chr.6: 32578904-32578905 on Build GRCh38
rs181125806 Chr.6: 32579137-32579137 on Build GRCh38
rs114103896 Chr.6: 32578782-32578782 on Build GRCh38
rs201085261 Chr.6: 32579227-32579227 on Build GRCh38
rs191464770 Chr.6: 32579240-32579240 on Build GRCh38
rs200601858 Chr.6: 32579111-32579111 on Build GRCh38
rs34864718 Chr.6: 32578765-32578765 on Build GRCh38
rs36084494 Chr.6: 32578996-32578996 on Build GRCh38
rs9269697 Chr.6: 32579176-32579176 on Build GRCh38
rs34844328 Chr.6: 32578997-32578997 on Build GRCh38
rs3179306 Chr.6: 32578950-32578950 on Build GRCh38
rs777165385 Chr.6: 32579126-32579126 on Build GRCh38
rs754612942 Chr.6: 32579020-32579020 on Build GRCh38
rs33962457 Chr.6: 32579228-32579228 on Build GRCh38
rs199851985 Chr.6: 32579175-32579175 on Build GRCh38
rs35463048 Chr.6: 32578982-32578982 on Build GRCh38
rs185448040 Chr.6: 32578842-32578842 on Build GRCh38
rs3201080 Chr.6: 32578858-32578858 on Build GRCh38
rs142402365 Chr.6: 32579212-32579212 on Build GRCh38
rs35418460 Chr.6: 32578846-32578846 on Build GRCh38
rs1064712 Chr.6: 32578896-32578896 on Build GRCh38
rs753402410 Chr.6: 32579022-32579023 on Build GRCh38
rs753021267 Chr.6: 32578983-32578983 on Build GRCh38
rs36024050 Chr.6: 32579119-32579119 on Build GRCh38
rs3180799 Chr.6: 32579025-32579025 on Build GRCh38
rs1064699 Chr.6: 32579029-32579029 on Build GRCh38
rs796250141 Chr.6: 32579227-32579228 on Build GRCh38
rs41287216 Chr.6: 32579188-32579188 on Build GRCh38
rs760049502 Chr.6: 32579075-32579076 on Build GRCh38
rs1064713 Chr.6: 32578889-32578889 on Build GRCh38
rs769982464 Chr.6: 32579012-32579012 on Build GRCh38
rs771348340 Chr.6: 32579078-32579079 on Build GRCh38
rs868048748 Chr.6: 32579095-32579095 on Build GRCh38
rs879558131 Chr.6: 32578814-32578815 on Build GRCh38
rs988340321 Chr.6: 32579195-32579195 on Build GRCh38
rs148582499 Chr.6: 32578959-32578960 on Build GRCh38
rs35306263 Chr.6: 32578907-32578908 on Build GRCh38
rs182030800 Chr.6: 32579006-32579006 on Build GRCh38
rs34252386 Chr.6: 32579135-32579136 on Build GRCh38
rs3201069 Chr.6: 32578888-32578888 on Build GRCh38
rs6920823 Chr.6: 32578845-32578845 on Build GRCh38
rs3208365 Chr.6: 32578814-32578814 on Build GRCh38
rs35287346 Chr.6: 32579242-32579242 on Build GRCh38
rs764735614 Chr.6: 32579028-32579029 on Build GRCh38
rs9269695 Chr.6: 32579164-32579164 on Build GRCh38
rs772066074 Chr.6: 32579009-32579009 on Build GRCh38
rs1064709 Chr.6: 32578921-32578921 on Build GRCh38
rs796306073 Chr.6: 32579023-32579026 on Build GRCh38
rs1064692 Chr.6: 32579062-32579062 on Build GRCh38
rs749335421 Chr.6: 32579050-32579050 on Build GRCh38
rs775540137 Chr.6: 32579013-32579013 on Build GRCh38
rs1064701 Chr.6: 32579018-32579018 on Build GRCh38
rs9269699 Chr.6: 32579227-32579227 on Build GRCh38
rs71535483 Chr.6: 32578814-32578815 on Build GRCh38
rs34955866 Chr.6: 32579086-32579086 on Build GRCh38
rs568633464 Chr.6: 32579216-32579216 on Build GRCh38
rs3208409 Chr.6: 32578919-32578919 on Build GRCh38
rs34402408 Chr.6: 32579237-32579237 on Build GRCh38
rs34160410 Chr.6: 32578962-32578963 on Build GRCh38
rs35513414 Chr.6: 32578823-32578823 on Build GRCh38
rs763280715 Chr.6: 32579014-32579014 on Build GRCh38
rs72847691 Chr.6: 32579163-32579163 on Build GRCh38
rs1064691 Chr.6: 32579069-32579069 on Build GRCh38
rs1060185 Chr.6: 32578907-32578907 on Build GRCh38
rs35086759 Chr.6: 32579226-32579226 on Build GRCh38
rs3200898 Chr.6: 32579015-32579015 on Build GRCh38
rs775883466 Chr.6: 32579110-32579111 on Build GRCh38
rs34374474 Chr.6: 32579162-32579162 on Build GRCh38
rs34022924 Chr.6: 32578988-32578988 on Build GRCh38
rs34009233 Chr.6: 32578976-32578976 on Build GRCh38
rs1064717 Chr.6: 32578838-32578838 on Build GRCh38
rs35868805 Chr.6: 32579166-32579166 on Build GRCh38
rs34981130 Chr.6: 32578893-32578893 on Build GRCh38
rs35985423 Chr.6: 32579117-32579117 on Build GRCh38
rs762554217 Chr.6: 32579142-32579142 on Build GRCh38
rs747981161 Chr.6: 32579008-32579008 on Build GRCh38
rs34748211 Chr.6: 32579219-32579220 on Build GRCh38
rs747939071 Chr.6: 32579060-32579061 on Build GRCh38
rs200538592 Chr.6: 32579059-32579059 on Build GRCh38
rs34475960 Chr.6: 32579099-32579099 on Build GRCh38
rs36042073 Chr.6: 32578821-32578821 on Build GRCh38
rs3200047 Chr.6: 32578922-32578922 on Build GRCh38
rs759675096 Chr.6: 32579011-32579011 on Build GRCh38
rs1141869 Chr.6: 32579017-32579017 on Build GRCh38
rs34261587 Chr.6: 32579238-32579238 on Build GRCh38
rs35413567 Chr.6: 32578822-32578822 on Build GRCh38
rs779563658 Chr.6: 32579002-32579002 on Build GRCh38
rs766819429 Chr.6: 32579040-32579040 on Build GRCh38
rs34453196 Chr.6: 32579232-32579232 on Build GRCh38
rs368419083 Chr.6: 32579026-32579027 on Build GRCh38
rs3180268 Chr.6: 32579026-32579026 on Build GRCh38
rs761293855 Chr.6: 32579038-32579038 on Build GRCh38
rs80136018 Chr.6: 32578877-32578877 on Build GRCh38
rs41287215 Chr.6: 32579171-32579171 on Build GRCh38
rs780993144 Chr.6: 32579048-32579048 on Build GRCh38
rs759691504 Chr.6: 32578970-32578970 on Build GRCh38
rs763867874 Chr.6: 32579075-32579075 on Build GRCh38
rs776837972 Chr.6: 32579109-32579109 on Build GRCh38
rs67419769 Chr.6: 32579224-32579224 on Build GRCh38
rs12363 Chr.6: 32579028-32579028 on Build GRCh38
rs1730 Chr.6: 32578897-32578897 on Build GRCh38
rs777757658 Chr.6: 32579046-32579046 on Build GRCh38
rs1064710 Chr.6: 32578920-32578920 on Build GRCh38
rs41287214 Chr.6: 32579161-32579161 on Build GRCh38
rs9269700 Chr.6: 32579243-32579243 on Build GRCh38
rs868362879 Chr.6: 32579098-32579098 on Build GRCh38
rs35705303 Chr.6: 32578816-32578816 on Build GRCh38
rs200849525 Chr.6: 32579047-32579047 on Build GRCh38
rs67916955 Chr.6: 32579228-32579228 on Build GRCh38
rs755638414 Chr.6: 32578998-32578998 on Build GRCh38
rs9269698 Chr.6: 32579209-32579209 on Build GRCh38
rs113493811 Chr.6: 32578902-32578903 on Build GRCh38
rs35014135 Chr.6: 32579174-32579174 on Build GRCh38
rs775216421 Chr.6: 32578925-32578925 on Build GRCh38
rs763779342 Chr.6: 32579143-32579143 on Build GRCh38
rs36217728 Chr.6: 32579055-32579055 on Build GRCh38
rs759703575 Chr.6: 32579109-32579110 on Build GRCh38
rs200428856 Chr.6: 32578832-32578839 on Build GRCh38
rs1064715 Chr.6: 32578849-32578849 on Build GRCh38
rs774571917 Chr.6: 32579032-32579032 on Build GRCh38
rs369671770 Chr.6: 32578813-32578814 on Build GRCh38
rs550953960 Chr.6: 32579235-32579235 on Build GRCh38
rs1729 Chr.6: 32578852-32578852 on Build GRCh38
rs1060270 Chr.6: 32578826-32578826 on Build GRCh38
rs67615547 Chr.6: 32579152-32579152 on Build GRCh38

View Full Product Details