Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32581473-32581970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DRB1
Entrez Gene ID
3123
Gene Name
major histocompatibility complex, class II, DR beta 1
Gene Aliases
DRB1, DRw10, HLA-DR1B, HLA-DRB, SS1
UniGene
Hs.534322
Gene Chromosome Location
Chr.6: 32578769-32589836 on Build GRCh38
Transcripts
NM_001243965.1 NM_002124.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4595096 c.562G>A p.V188M Chr.6: 32581647-32581647 on Build GRCh38
4160962 c.511A>G p.M171V Chr.6: 32581698-32581698 on Build GRCh38
3733801 c.397T>G p.S133A Chr.6: 32581812-32581812 on Build GRCh38
106766 c.394C>T p.P132S Chr.6: 32581815-32581815 on Build GRCh38
3830223 c.423C>T p.H141H Chr.6: 32581786-32581786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753240578 Chr.6: 32581784-32581784 on Build GRCh38
rs772667813 Chr.6: 32581869-32581869 on Build GRCh38
rs760207412 Chr.6: 32581857-32581857 on Build GRCh38
rs372360291 Chr.6: 32581875-32581875 on Build GRCh38
rs757966595 Chr.6: 32581719-32581719 on Build GRCh38
rs747401954 Chr.6: 32581508-32581508 on Build GRCh38
rs201198935 Chr.6: 32581528-32581528 on Build GRCh38
rs701831 Chr.6: 32581624-32581624 on Build GRCh38
rs74867190 Chr.6: 32581666-32581666 on Build GRCh38
rs78916069 Chr.6: 32581704-32581704 on Build GRCh38
rs750301093 Chr.6: 32581878-32581878 on Build GRCh38
rs775763997 Chr.6: 32581845-32581845 on Build GRCh38
rs9269798 Chr.6: 32581519-32581519 on Build GRCh38
rs759467362 Chr.6: 32581665-32581665 on Build GRCh38
rs781450663 Chr.6: 32581842-32581842 on Build GRCh38
rs9269796 Chr.6: 32581501-32581501 on Build GRCh38
rs16822698 Chr.6: 32581748-32581748 on Build GRCh38
rs878955279 Chr.6: 32581800-32581800 on Build GRCh38
rs759833767 Chr.6: 32581776-32581776 on Build GRCh38
rs748235111 Chr.6: 32581752-32581752 on Build GRCh38
rs17885192 Chr.6: 32581774-32581774 on Build GRCh38
rs199923212 Chr.6: 32581520-32581520 on Build GRCh38
rs28732327 Chr.6: 32581498-32581498 on Build GRCh38
rs201929247 Chr.6: 32581811-32581811 on Build GRCh38
rs763319916 Chr.6: 32581847-32581847 on Build GRCh38
rs778232030 Chr.6: 32581850-32581851 on Build GRCh38
rs35172954 Chr.6: 32581522-32581522 on Build GRCh38
rs28732338 Chr.6: 32581925-32581925 on Build GRCh38
rs9269814 Chr.6: 32581934-32581934 on Build GRCh38
rs767415337 Chr.6: 32581854-32581855 on Build GRCh38
rs771739125 Chr.6: 32581600-32581600 on Build GRCh38
rs763899183 Chr.6: 32581851-32581854 on Build GRCh38
rs62799161 Chr.6: 32581831-32581831 on Build GRCh38
rs199927008 Chr.6: 32581850-32581850 on Build GRCh38
rs879790499 Chr.6: 32581718-32581718 on Build GRCh38
rs769469262 Chr.6: 32581511-32581511 on Build GRCh38
rs781648698 Chr.6: 32581581-32581581 on Build GRCh38
rs759188166 Chr.6: 32581850-32581862 on Build GRCh38
rs2308769 Chr.6: 32581692-32581692 on Build GRCh38
rs1136846 Chr.6: 32581575-32581575 on Build GRCh38
rs111739605 Chr.6: 32581584-32581584 on Build GRCh38
rs16822692 Chr.6: 32581639-32581639 on Build GRCh38
rs62930562 Chr.6: 32581660-32581660 on Build GRCh38
rs756424707 Chr.6: 32581793-32581793 on Build GRCh38
rs762530337 Chr.6: 32581851-32581856 on Build GRCh38
rs34938122 Chr.6: 32581603-32581603 on Build GRCh38
rs753045406 Chr.6: 32581803-32581803 on Build GRCh38
rs35493155 Chr.6: 32581510-32581510 on Build GRCh38
rs754428084 Chr.6: 32581626-32581626 on Build GRCh38
rs879752051 Chr.6: 32581813-32581813 on Build GRCh38
rs9269799 Chr.6: 32581536-32581536 on Build GRCh38
rs3205588 Chr.6: 32581625-32581625 on Build GRCh38
rs35464026 Chr.6: 32581533-32581533 on Build GRCh38
rs775098518 Chr.6: 32581542-32581542 on Build GRCh38
rs9269797 Chr.6: 32581507-32581507 on Build GRCh38
rs143082263 Chr.6: 32581865-32581865 on Build GRCh38
rs1059615 Chr.6: 32581780-32581780 on Build GRCh38
rs776695055 Chr.6: 32581614-32581614 on Build GRCh38
rs148699775 Chr.6: 32581815-32581815 on Build GRCh38
rs761090565 Chr.6: 32581690-32581690 on Build GRCh38
rs1071752 Chr.6: 32581834-32581834 on Build GRCh38
rs879184569 Chr.6: 32581678-32581678 on Build GRCh38
rs375443740 Chr.6: 32581851-32581852 on Build GRCh38
rs765354312 Chr.6: 32581521-32581521 on Build GRCh38
rs34426449 Chr.6: 32581512-32581512 on Build GRCh38
rs17424180 Chr.6: 32581828-32581828 on Build GRCh38
rs17405325 Chr.6: 32581606-32581606 on Build GRCh38
rs701829 Chr.6: 32581698-32581698 on Build GRCh38
rs9269813 Chr.6: 32581904-32581904 on Build GRCh38
rs535956089 Chr.6: 32581906-32581906 on Build GRCh38
rs754180747 Chr.6: 32581703-32581703 on Build GRCh38
rs4999094 Chr.6: 32581848-32581848 on Build GRCh38
rs757298489 Chr.6: 32581854-32581854 on Build GRCh38
rs34295373 Chr.6: 32581591-32581591 on Build GRCh38
rs41293159 Chr.6: 32581886-32581886 on Build GRCh38
rs35924155 Chr.6: 32581509-32581509 on Build GRCh38
rs2308771 Chr.6: 32581669-32581669 on Build GRCh38
rs28732339 Chr.6: 32581941-32581941 on Build GRCh38
rs4999093 Chr.6: 32581851-32581851 on Build GRCh38
rs779011813 Chr.6: 32581839-32581839 on Build GRCh38
rs113663708 Chr.6: 32581651-32581651 on Build GRCh38
rs772406969 Chr.6: 32581849-32581851 on Build GRCh38
rs17885501 Chr.6: 32581574-32581574 on Build GRCh38
rs707941 Chr.6: 32581771-32581771 on Build GRCh38
rs9269811 Chr.6: 32581872-32581872 on Build GRCh38
rs760934639 Chr.6: 32581761-32581761 on Build GRCh38
rs770952933 Chr.6: 32581840-32581840 on Build GRCh38
rs28732340 Chr.6: 32581945-32581945 on Build GRCh38
rs9269815 Chr.6: 32581949-32581949 on Build GRCh38
rs202047044 Chr.6: 32581743-32581743 on Build GRCh38
rs1064596 Chr.6: 32581798-32581798 on Build GRCh38
rs41557115 Chr.6: 32581782-32581782 on Build GRCh38
rs2308775 Chr.6: 32581619-32581619 on Build GRCh38
rs1059705 Chr.6: 32581582-32581582 on Build GRCh38
rs76895951 Chr.6: 32581823-32581823 on Build GRCh38
rs34944636 Chr.6: 32581874-32581874 on Build GRCh38
rs768974536 Chr.6: 32581845-32581845 on Build GRCh38
rs17433947 Chr.6: 32581806-32581806 on Build GRCh38
rs79706935 Chr.6: 32581832-32581832 on Build GRCh38
rs760278421 Chr.6: 32581870-32581870 on Build GRCh38
rs778993385 Chr.6: 32581860-32581860 on Build GRCh38
rs746056914 Chr.6: 32581783-32581785 on Build GRCh38
rs28732335 Chr.6: 32581900-32581900 on Build GRCh38
rs707954 Chr.6: 32581724-32581724 on Build GRCh38
rs750742941 Chr.6: 32581688-32581688 on Build GRCh38
rs759378918 Chr.6: 32581873-32581873 on Build GRCh38
rs757139064 Chr.6: 32581622-32581622 on Build GRCh38
rs72114462 Chr.6: 32581851-32581851 on Build GRCh38
rs16822972 Chr.6: 32581615-32581615 on Build GRCh38
rs761684252 Chr.6: 32581516-32581516 on Build GRCh38
rs36056209 Chr.6: 32581859-32581859 on Build GRCh38
rs2308818 Chr.6: 32581579-32581579 on Build GRCh38
rs748466756 Chr.6: 32581845-32581846 on Build GRCh38
rs773389640 Chr.6: 32581821-32581821 on Build GRCh38
rs34363403 Chr.6: 32581866-32581866 on Build GRCh38
rs2308770 Chr.6: 32581681-32581681 on Build GRCh38
rs4999092 Chr.6: 32581854-32581854 on Build GRCh38
rs774111408 Chr.6: 32581549-32581549 on Build GRCh38
rs34450868 Chr.6: 32581506-32581506 on Build GRCh38
rs752658977 Chr.6: 32581568-32581568 on Build GRCh38
rs41292734 Chr.6: 32581540-32581540 on Build GRCh38
rs200088269 Chr.6: 32581807-32581808 on Build GRCh38
rs774805695 Chr.6: 32581851-32581858 on Build GRCh38
rs769138784 Chr.6: 32581852-32581852 on Build GRCh38
rs375154056 Chr.6: 32581625-32581626 on Build GRCh38
rs756774358 Chr.6: 32581835-32581835 on Build GRCh38
rs2308759 Chr.6: 32581819-32581819 on Build GRCh38
rs28732334 Chr.6: 32581898-32581898 on Build GRCh38
rs77637983 Chr.6: 32581675-32581675 on Build GRCh38
rs33936261 Chr.6: 32581563-32581563 on Build GRCh38
rs33926824 Chr.6: 32581578-32581578 on Build GRCh38
rs67184706 Chr.6: 32581846-32581846 on Build GRCh38
rs2308777 Chr.6: 32581609-32581609 on Build GRCh38
rs72850244 Chr.6: 32581861-32581861 on Build GRCh38
rs879379639 Chr.6: 32581714-32581714 on Build GRCh38
rs747689830 Chr.6: 32581634-32581634 on Build GRCh38
rs754898862 Chr.6: 32581838-32581838 on Build GRCh38
rs35129150 Chr.6: 32581892-32581892 on Build GRCh38
rs572938448 Chr.6: 32581673-32581673 on Build GRCh38
rs750453829 Chr.6: 32581887-32581887 on Build GRCh38
rs751612214 Chr.6: 32581618-32581618 on Build GRCh38
rs9269812 Chr.6: 32581893-32581893 on Build GRCh38
rs758200279 Chr.6: 32581885-32581885 on Build GRCh38
rs200516145 Chr.6: 32581805-32581805 on Build GRCh38
rs1136791 Chr.6: 32581824-32581824 on Build GRCh38
rs751099504 Chr.6: 32581788-32581788 on Build GRCh38
rs35615823 Chr.6: 32581504-32581504 on Build GRCh38
rs779295390 Chr.6: 32581711-32581711 on Build GRCh38
rs140866337 Chr.6: 32581837-32581837 on Build GRCh38
rs35616319 Chr.6: 32581810-32581811 on Build GRCh38
rs145408477 Chr.6: 32581513-32581513 on Build GRCh38
rs34382076 Chr.6: 32581548-32581548 on Build GRCh38
rs1071837 Chr.6: 32581573-32581573 on Build GRCh38
rs771131230 Chr.6: 32581827-32581827 on Build GRCh38
rs35527347 Chr.6: 32581514-32581514 on Build GRCh38
rs554801069 Chr.6: 32581946-32581946 on Build GRCh38
rs2308761 Chr.6: 32581810-32581810 on Build GRCh38
rs80190494 Chr.6: 32581826-32581826 on Build GRCh38
rs2308767 Chr.6: 32581720-32581720 on Build GRCh38
rs774787256 Chr.6: 32581853-32581853 on Build GRCh38
rs36044702 Chr.6: 32581599-32581599 on Build GRCh38
rs78767604 Chr.6: 32581687-32581687 on Build GRCh38
rs140357311 Chr.6: 32581597-32581616 on Build GRCh38
rs879030354 Chr.6: 32581808-32581808 on Build GRCh38
rs778066941 Chr.6: 32581525-32581525 on Build GRCh38
rs745655219 Chr.6: 32581735-32581735 on Build GRCh38
rs745962012 Chr.6: 32581862-32581862 on Build GRCh38
rs111760370 Chr.6: 32581502-32581502 on Build GRCh38
rs2308765 Chr.6: 32581757-32581757 on Build GRCh38
rs17405219 Chr.6: 32581830-32581830 on Build GRCh38
rs746709087 Chr.6: 32581723-32581723 on Build GRCh38
rs3200488 Chr.6: 32581585-32581585 on Build GRCh38
rs62889561 Chr.6: 32581822-32581822 on Build GRCh38
rs112116022 Chr.6: 32581647-32581647 on Build GRCh38
rs761509315 Chr.6: 32581551-32581551 on Build GRCh38
rs751518272 Chr.6: 32581852-32581853 on Build GRCh38
rs768278955 Chr.6: 32581586-32581586 on Build GRCh38
rs200809587 Chr.6: 32581580-32581580 on Build GRCh38
rs200078051 Chr.6: 32581807-32581807 on Build GRCh38
rs752147302 Chr.6: 32581717-32581717 on Build GRCh38
rs74944677 Chr.6: 32581825-32581825 on Build GRCh38
rs28732336 Chr.6: 32581902-32581902 on Build GRCh38
rs34837048 Chr.6: 32581503-32581503 on Build GRCh38
rs17401330 Chr.6: 32581804-32581804 on Build GRCh38
rs28732337 Chr.6: 32581919-32581919 on Build GRCh38
rs766136796 Chr.6: 32581858-32581858 on Build GRCh38
rs34309628 Chr.6: 32581554-32581554 on Build GRCh38
rs778441312 Chr.6: 32581633-32581633 on Build GRCh38
rs35837054 Chr.6: 32581568-32581569 on Build GRCh38
rs199824918 Chr.6: 32581697-32581699 on Build GRCh38
rs201219028 Chr.6: 32581843-32581843 on Build GRCh38
rs770748207 Chr.6: 32581653-32581653 on Build GRCh38
rs35300013 Chr.6: 32581622-32581622 on Build GRCh38
rs2308760 Chr.6: 32581812-32581812 on Build GRCh38
rs63548860 Chr.6: 32581696-32581696 on Build GRCh38
rs1059351 Chr.6: 32581763-32581763 on Build GRCh38
rs77689370 Chr.6: 32581786-32581786 on Build GRCh38
rs748834188 Chr.6: 32581543-32581543 on Build GRCh38
rs17882084 Chr.6: 32581836-32581836 on Build GRCh38
rs112796209 Chr.6: 32581754-32581754 on Build GRCh38
rs752565649 Chr.6: 32581877-32581877 on Build GRCh38
rs879211739 Chr.6: 32581579-32581580 on Build GRCh38
rs41293164 Chr.6: 32581890-32581890 on Build GRCh38
rs774901145 Chr.6: 32581868-32581868 on Build GRCh38
rs17878677 Chr.6: 32581567-32581567 on Build GRCh38
rs758511696 Chr.6: 32581523-32581523 on Build GRCh38
rs879337960 Chr.6: 32581765-32581765 on Build GRCh38
rs747318806 Chr.6: 32581527-32581527 on Build GRCh38
rs36070756 Chr.6: 32581517-32581517 on Build GRCh38
rs773547095 Chr.6: 32581756-32581756 on Build GRCh38
rs200922520 Chr.6: 32581515-32581515 on Build GRCh38

View Full Product Details