Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32584321-32584736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DRB1
Entrez Gene ID
3123
Gene Name
major histocompatibility complex, class II, DR beta 1
Gene Aliases
DRB1, DRw10, HLA-DR1B, HLA-DRB, SS1
UniGene
Hs.534322
Gene Chromosome Location
Chr.6: 32578769-32589836 on Build GRCh38
Transcripts
NM_001243965.1 NM_002124.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4160978 c.109C>T p.L37L Chr.6: 32584370-32584370 on Build GRCh38
4594955 c.125G>C p.R42T Chr.6: 32584354-32584354 on Build GRCh38
4160977 c.112T>G p.W38G Chr.6: 32584367-32584367 on Build GRCh38
3722067 c.144T>C p.N48N Chr.6: 32584335-32584335 on Build GRCh38
451270 c.101-1G>A p.? Chr.6: 32584379-32584379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs28724119 Chr.6: 32584527-32584527 on Build GRCh38
rs114610112 Chr.6: 32584382-32584382 on Build GRCh38
rs759539540 Chr.6: 32584493-32584493 on Build GRCh38
rs36170912 Chr.6: 32584607-32584608 on Build GRCh38
rs201335665 Chr.6: 32584600-32584600 on Build GRCh38
rs765291187 Chr.6: 32584554-32584554 on Build GRCh38
rs774455138 Chr.6: 32584364-32584366 on Build GRCh38
rs28724115 Chr.6: 32584510-32584510 on Build GRCh38
rs147863572 Chr.6: 32584645-32584645 on Build GRCh38
rs751151553 Chr.6: 32584395-32584396 on Build GRCh38
rs773489989 Chr.6: 32584368-32584368 on Build GRCh38
rs41287315 Chr.6: 32584627-32584627 on Build GRCh38
rs772011591 Chr.6: 32584366-32584367 on Build GRCh38
rs776465322 Chr.6: 32584350-32584351 on Build GRCh38
rs9269960 Chr.6: 32584370-32584370 on Build GRCh38
rs775308685 Chr.6: 32584379-32584379 on Build GRCh38
rs1009685936 Chr.6: 32584512-32584512 on Build GRCh38
rs770838956 Chr.6: 32584350-32584353 on Build GRCh38
rs28724110 Chr.6: 32584424-32584424 on Build GRCh38
rs28724105 Chr.6: 32584391-32584391 on Build GRCh38
rs757711936 Chr.6: 32584410-32584410 on Build GRCh38
rs9269979 Chr.6: 32584589-32584589 on Build GRCh38
rs9269976 Chr.6: 32584571-32584571 on Build GRCh38
rs28986202 Chr.6: 32584409-32584410 on Build GRCh38
rs28724112 Chr.6: 32584485-32584485 on Build GRCh38
rs762139928 Chr.6: 32584365-32584366 on Build GRCh38
rs538710337 Chr.6: 32584614-32584614 on Build GRCh38
rs28724109 Chr.6: 32584421-32584421 on Build GRCh38
rs17882014 Chr.6: 32584352-32584352 on Build GRCh38
rs9269969 Chr.6: 32584495-32584495 on Build GRCh38
rs41287319 Chr.6: 32584686-32584686 on Build GRCh38
rs1136756 Chr.6: 32584357-32584357 on Build GRCh38
rs28724130 Chr.6: 32584711-32584711 on Build GRCh38
rs41285659 Chr.6: 32584396-32584396 on Build GRCh38
rs745775563 Chr.6: 32584358-32584359 on Build GRCh38
rs149614319 Chr.6: 32584446-32584446 on Build GRCh38
rs9269971 Chr.6: 32584514-32584514 on Build GRCh38
rs370480631 Chr.6: 32584407-32584408 on Build GRCh38
rs942975942 Chr.6: 32584563-32584563 on Build GRCh38
rs766505678 Chr.6: 32584358-32584358 on Build GRCh38
rs796995514 Chr.6: 32584583-32584585 on Build GRCh38
rs774339131 Chr.6: 32584388-32584388 on Build GRCh38
rs28724126 Chr.6: 32584628-32584628 on Build GRCh38
rs41287323 Chr.6: 32584689-32584689 on Build GRCh38
rs9269964 Chr.6: 32584435-32584435 on Build GRCh38
rs36178790 Chr.6: 32584653-32584653 on Build GRCh38
rs28724114 Chr.6: 32584507-32584507 on Build GRCh38
rs143441545 Chr.6: 32584488-32584488 on Build GRCh38
rs28724129 Chr.6: 32584690-32584690 on Build GRCh38
rs758070868 Chr.6: 32584355-32584356 on Build GRCh38
rs769556955 Chr.6: 32584352-32584353 on Build GRCh38
rs28724104 Chr.6: 32584387-32584387 on Build GRCh38
rs9269956 Chr.6: 32584362-32584362 on Build GRCh38
rs756741350 Chr.6: 32584357-32584361 on Build GRCh38
rs112353158 Chr.6: 32584411-32584411 on Build GRCh38
rs1136758 Chr.6: 32584355-32584355 on Build GRCh38
rs17885011 Chr.6: 32584350-32584350 on Build GRCh38
rs41288201 Chr.6: 32584708-32584709 on Build GRCh38
rs28724118 Chr.6: 32584519-32584519 on Build GRCh38
rs9269970 Chr.6: 32584496-32584496 on Build GRCh38
rs140806391 Chr.6: 32584406-32584407 on Build GRCh38
rs759853937 Chr.6: 32584393-32584394 on Build GRCh38
rs760729552 Chr.6: 32584390-32584390 on Build GRCh38
rs34951355 Chr.6: 32584639-32584639 on Build GRCh38
rs768015363 Chr.6: 32584353-32584354 on Build GRCh38
rs115462544 Chr.6: 32584452-32584452 on Build GRCh38
rs766848979 Chr.6: 32584355-32584361 on Build GRCh38
rs28724125 Chr.6: 32584582-32584582 on Build GRCh38
rs548073851 Chr.6: 32584580-32584580 on Build GRCh38
rs9256941 Chr.6: 32584407-32584407 on Build GRCh38
rs71540455 Chr.6: 32584592-32584592 on Build GRCh38
rs375544035 Chr.6: 32584712-32584712 on Build GRCh38
rs779612952 Chr.6: 32584415-32584415 on Build GRCh38
rs144353907 Chr.6: 32584449-32584449 on Build GRCh38
rs17879981 Chr.6: 32584344-32584344 on Build GRCh38
rs115170333 Chr.6: 32584545-32584545 on Build GRCh38
rs376098112 Chr.6: 32584603-32584604 on Build GRCh38
rs11753207 Chr.6: 32584671-32584671 on Build GRCh38
rs116085021 Chr.6: 32584594-32584594 on Build GRCh38
rs17878703 Chr.6: 32584360-32584360 on Build GRCh38
rs9269980 Chr.6: 32584618-32584618 on Build GRCh38
rs373637765 Chr.6: 32584635-32584635 on Build GRCh38
rs376484792 Chr.6: 32584678-32584678 on Build GRCh38
rs28724107 Chr.6: 32584418-32584418 on Build GRCh38
rs796516603 Chr.6: 32584688-32584688 on Build GRCh38
rs9269972 Chr.6: 32584525-32584525 on Build GRCh38
rs28724108 Chr.6: 32584420-32584420 on Build GRCh38
rs754316114 Chr.6: 32584356-32584360 on Build GRCh38
rs186922906 Chr.6: 32584595-32584595 on Build GRCh38
rs28724121 Chr.6: 32584547-32584547 on Build GRCh38
rs9269957 Chr.6: 32584364-32584364 on Build GRCh38
rs746487937 Chr.6: 32584356-32584357 on Build GRCh38
rs776415590 Chr.6: 32584343-32584343 on Build GRCh38
rs28724128 Chr.6: 32584668-32584668 on Build GRCh38
rs754191737 Chr.6: 32584383-32584383 on Build GRCh38
rs28724116 Chr.6: 32584511-32584511 on Build GRCh38
rs28724123 Chr.6: 32584559-32584559 on Build GRCh38
rs531110373 Chr.6: 32584470-32584470 on Build GRCh38
rs745460990 Chr.6: 32584378-32584378 on Build GRCh38
rs774423362 Chr.6: 32584353-32584358 on Build GRCh38
rs761252314 Chr.6: 32584368-32584369 on Build GRCh38
rs9269973 Chr.6: 32584530-32584530 on Build GRCh38
rs9269974 Chr.6: 32584536-32584536 on Build GRCh38
rs756975747 Chr.6: 32584402-32584403 on Build GRCh38
rs974401591 Chr.6: 32584675-32584675 on Build GRCh38
rs9269963 Chr.6: 32584429-32584429 on Build GRCh38
rs752059618 Chr.6: 32584408-32584408 on Build GRCh38
rs1136759 Chr.6: 32584354-32584354 on Build GRCh38
rs369093408 Chr.6: 32584679-32584679 on Build GRCh38
rs749085224 Chr.6: 32584361-32584362 on Build GRCh38
rs9269951 Chr.6: 32584353-32584353 on Build GRCh38
rs9269967 Chr.6: 32584462-32584462 on Build GRCh38
rs9269978 Chr.6: 32584585-32584585 on Build GRCh38
rs9281875 Chr.6: 32584612-32584613 on Build GRCh38
rs41285655 Chr.6: 32584394-32584394 on Build GRCh38
rs28724113 Chr.6: 32584504-32584504 on Build GRCh38
rs149553038 Chr.6: 32584476-32584477 on Build GRCh38
rs796252808 Chr.6: 32584706-32584706 on Build GRCh38
rs9269962 Chr.6: 32584400-32584400 on Build GRCh38
rs749029126 Chr.6: 32584428-32584456 on Build GRCh38
rs36177427 Chr.6: 32584684-32584684 on Build GRCh38
rs41285663 Chr.6: 32584454-32584454 on Build GRCh38
rs9269975 Chr.6: 32584553-32584553 on Build GRCh38
rs17879702 Chr.6: 32584346-32584346 on Build GRCh38
rs17887028 Chr.6: 32584356-32584356 on Build GRCh38
rs768883657 Chr.6: 32584364-32584364 on Build GRCh38
rs769779152 Chr.6: 32584360-32584361 on Build GRCh38
rs28724122 Chr.6: 32584551-32584551 on Build GRCh38
rs28724111 Chr.6: 32584463-32584463 on Build GRCh38
rs534288394 Chr.6: 32584685-32584685 on Build GRCh38
rs767010367 Chr.6: 32584370-32584370 on Build GRCh38
rs9269966 Chr.6: 32584460-32584460 on Build GRCh38
rs9269961 Chr.6: 32584395-32584395 on Build GRCh38
rs139860564 Chr.6: 32584699-32584699 on Build GRCh38
rs28724117 Chr.6: 32584513-32584513 on Build GRCh38
rs41285667 Chr.6: 32584539-32584539 on Build GRCh38
rs28724124 Chr.6: 32584573-32584573 on Build GRCh38
rs9269955 Chr.6: 32584361-32584361 on Build GRCh38
rs9269958 Chr.6: 32584366-32584366 on Build GRCh38
rs547038817 Chr.6: 32584520-32584520 on Build GRCh38
rs28724106 Chr.6: 32584413-32584413 on Build GRCh38
rs28724127 Chr.6: 32584643-32584643 on Build GRCh38
rs763753913 Chr.6: 32584394-32584399 on Build GRCh38
rs372051739 Chr.6: 32584711-32584711 on Build GRCh38
rs41285671 Chr.6: 32584569-32584569 on Build GRCh38
rs796438888 Chr.6: 32584460-32584462 on Build GRCh38
rs9269965 Chr.6: 32584442-32584442 on Build GRCh38
rs566656642 Chr.6: 32584562-32584562 on Build GRCh38
rs17882663 Chr.6: 32584363-32584363 on Build GRCh38
rs9281876 Chr.6: 32584705-32584706 on Build GRCh38
rs796657099 Chr.6: 32584696-32584696 on Build GRCh38
rs373304672 Chr.6: 32584703-32584704 on Build GRCh38
rs796508124 Chr.6: 32584707-32584707 on Build GRCh38
rs9281874 Chr.6: 32584478-32584479 on Build GRCh38
rs190180919 Chr.6: 32584386-32584386 on Build GRCh38
rs745615811 Chr.6: 32584352-32584354 on Build GRCh38
rs9269968 Chr.6: 32584474-32584474 on Build GRCh38
rs41287310 Chr.6: 32584577-32584577 on Build GRCh38
rs28724120 Chr.6: 32584542-32584542 on Build GRCh38
rs780805378 Chr.6: 32584357-32584358 on Build GRCh38
rs753085642 Chr.6: 32584403-32584403 on Build GRCh38
rs41308497 Chr.6: 32584376-32584376 on Build GRCh38
rs114435757 Chr.6: 32584340-32584340 on Build GRCh38
rs9269977 Chr.6: 32584583-32584583 on Build GRCh38
rs9269959 Chr.6: 32584367-32584367 on Build GRCh38

View Full Product Details