Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32659755-32660290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DQB1
Entrez Gene ID
3119
Gene Name
major histocompatibility complex, class II, DQ beta 1
Gene Aliases
CELIAC1, HLA-DQB, HLA-DRB1, IDDM1
UniGene
Hs.409934
Gene Chromosome Location
Chr.6: 32659464-32666689 on Build GRCh38
Transcripts
NM_001243961.1 NM_001243962.1 NM_002123.4 NR_133907.1

Gene Symbol
HLA-DQB1-AS1
Entrez Gene ID
106480429
Gene Name
HLA-DQB1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.701274
Gene Chromosome Location
Chr.6: 32659880-32660729 on Build GRCh38
Transcripts
NM_001243961.1 NM_001243962.1 NM_002123.4 NR_133907.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1621489 c.777T>C p.L259L Chr.6: 32660245-32660245 on Build GRCh38
1621490 c.773-4C>T p.? Chr.6: 32660253-32660253 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1130422 Chr.6: 32660067-32660067 on Build GRCh38
rs988508070 Chr.6: 32659894-32659894 on Build GRCh38
rs1049124 Chr.6: 32659936-32659936 on Build GRCh38
rs7656 Chr.6: 32660181-32660181 on Build GRCh38
rs1049178 Chr.6: 32660211-32660211 on Build GRCh38
rs1049225 Chr.6: 32659970-32659970 on Build GRCh38
rs113664950 Chr.6: 32660108-32660108 on Build GRCh38
rs3842708 Chr.6: 32660137-32660137 on Build GRCh38
rs117789582 Chr.6: 32659826-32659826 on Build GRCh38
rs9273440 Chr.6: 32659784-32659784 on Build GRCh38
rs183950119 Chr.6: 32659981-32659981 on Build GRCh38
rs201738242 Chr.6: 32660202-32660202 on Build GRCh38
rs9273444 Chr.6: 32659858-32659858 on Build GRCh38
rs774067936 Chr.6: 32660268-32660270 on Build GRCh38
rs1049213 Chr.6: 32659996-32659996 on Build GRCh38
rs765136994 Chr.6: 32660188-32660188 on Build GRCh38
rs1071639 Chr.6: 32660105-32660105 on Build GRCh38
rs1049123 Chr.6: 32660017-32660017 on Build GRCh38
rs766570827 Chr.6: 32660197-32660197 on Build GRCh38
rs756361095 Chr.6: 32659911-32659912 on Build GRCh38
rs914104471 Chr.6: 32659909-32659909 on Build GRCh38
rs1770 Chr.6: 32660056-32660056 on Build GRCh38
rs943602050 Chr.6: 32659951-32659951 on Build GRCh38
rs1140365 Chr.6: 32659866-32659866 on Build GRCh38
rs6689 Chr.6: 32659923-32659923 on Build GRCh38
rs771958616 Chr.6: 32660136-32660137 on Build GRCh38
rs1063346 Chr.6: 32660190-32660190 on Build GRCh38
rs952726949 Chr.6: 32659781-32659781 on Build GRCh38
rs910800854 Chr.6: 32659944-32659944 on Build GRCh38
rs796842827 Chr.6: 32660066-32660067 on Build GRCh38
rs545181996 Chr.6: 32659806-32659806 on Build GRCh38
rs9273463 Chr.6: 32660157-32660157 on Build GRCh38
rs9273445 Chr.6: 32659869-32659869 on Build GRCh38
rs1014513276 Chr.6: 32660109-32660109 on Build GRCh38
rs1049122 Chr.6: 32660030-32660030 on Build GRCh38
rs755264941 Chr.6: 32660206-32660206 on Build GRCh38
rs1049219 Chr.6: 32659986-32659986 on Build GRCh38
rs4993988 Chr.6: 32660007-32660007 on Build GRCh38
rs930155204 Chr.6: 32660013-32660013 on Build GRCh38
rs756357005 Chr.6: 32660195-32660195 on Build GRCh38
rs1130446 Chr.6: 32659840-32659840 on Build GRCh38
rs3204435 Chr.6: 32659856-32659856 on Build GRCh38
rs910854110 Chr.6: 32659997-32659997 on Build GRCh38
rs1762 Chr.6: 32660000-32660000 on Build GRCh38
rs1049177 Chr.6: 32660214-32660214 on Build GRCh38
rs386699568 Chr.6: 32660129-32660146 on Build GRCh38
rs181293540 Chr.6: 32660043-32660043 on Build GRCh38
rs1140347 Chr.6: 32660245-32660245 on Build GRCh38
rs535153974 Chr.6: 32660134-32660134 on Build GRCh38
rs9273453 Chr.6: 32660035-32660035 on Build GRCh38
rs879601773 Chr.6: 32660074-32660075 on Build GRCh38
rs970389732 Chr.6: 32659943-32659943 on Build GRCh38
rs3210187 Chr.6: 32659889-32659889 on Build GRCh38
rs1049222 Chr.6: 32659975-32659975 on Build GRCh38
rs9273471 Chr.6: 32660253-32660253 on Build GRCh38
rs978411868 Chr.6: 32659861-32659861 on Build GRCh38
rs3204433 Chr.6: 32659891-32659891 on Build GRCh38
rs9282076 Chr.6: 32660068-32660069 on Build GRCh38
rs35888564 Chr.6: 32660145-32660145 on Build GRCh38
rs115749644 Chr.6: 32660041-32660041 on Build GRCh38
rs34153824 Chr.6: 32659850-32659850 on Build GRCh38
rs929412296 Chr.6: 32660063-32660063 on Build GRCh38
rs1063343 Chr.6: 32660215-32660215 on Build GRCh38
rs1063345 Chr.6: 32660199-32660199 on Build GRCh38
rs1768 Chr.6: 32660041-32660041 on Build GRCh38
rs9273441 Chr.6: 32659796-32659796 on Build GRCh38
rs750510294 Chr.6: 32660194-32660194 on Build GRCh38
rs947115310 Chr.6: 32659989-32659989 on Build GRCh38
rs748609904 Chr.6: 32660209-32660209 on Build GRCh38
rs1063348 Chr.6: 32660146-32660146 on Build GRCh38
rs764003327 Chr.6: 32660267-32660267 on Build GRCh38
rs9273459 Chr.6: 32660074-32660074 on Build GRCh38
rs1766 Chr.6: 32660018-32660018 on Build GRCh38
rs866897949 Chr.6: 32660269-32660269 on Build GRCh38
rs994333262 Chr.6: 32659883-32659883 on Build GRCh38
rs3210176 Chr.6: 32660073-32660073 on Build GRCh38
rs1063355 Chr.6: 32659937-32659937 on Build GRCh38
rs775501650 Chr.6: 32660189-32660189 on Build GRCh38
rs9273472 Chr.6: 32660268-32660268 on Build GRCh38
rs796318446 Chr.6: 32660190-32660192 on Build GRCh38
rs1071641 Chr.6: 32660021-32660021 on Build GRCh38
rs386699570 Chr.6: 32660268-32660270 on Build GRCh38
rs1130445 Chr.6: 32659843-32659843 on Build GRCh38
rs759460475 Chr.6: 32660186-32660186 on Build GRCh38
rs888653196 Chr.6: 32660162-32660162 on Build GRCh38
rs1130420 Chr.6: 32660075-32660075 on Build GRCh38
rs1764 Chr.6: 32660008-32660008 on Build GRCh38
rs1021374283 Chr.6: 32660068-32660068 on Build GRCh38
rs4993986 Chr.6: 32659875-32659875 on Build GRCh38
rs139850766 Chr.6: 32660225-32660225 on Build GRCh38
rs762916669 Chr.6: 32660266-32660266 on Build GRCh38
rs36205798 Chr.6: 32659836-32659836 on Build GRCh38
rs914304936 Chr.6: 32659842-32659842 on Build GRCh38
rs779360470 Chr.6: 32660140-32660140 on Build GRCh38
rs9273455 Chr.6: 32660053-32660053 on Build GRCh38
rs1049175 Chr.6: 32660224-32660224 on Build GRCh38
rs776479724 Chr.6: 32660185-32660185 on Build GRCh38
rs117067420 Chr.6: 32659877-32659877 on Build GRCh38
rs779276671 Chr.6: 32660208-32660208 on Build GRCh38
rs9273443 Chr.6: 32659823-32659823 on Build GRCh38
rs1036016695 Chr.6: 32659972-32659972 on Build GRCh38
rs1024133287 Chr.6: 32660033-32660033 on Build GRCh38
rs386699569 Chr.6: 32660179-32660181 on Build GRCh38
rs281864466 Chr.6: 32660067-32660067 on Build GRCh38
rs1063349 Chr.6: 32660129-32660129 on Build GRCh38
rs9273442 Chr.6: 32659805-32659805 on Build GRCh38
rs776366076 Chr.6: 32660241-32660241 on Build GRCh38
rs1063347 Chr.6: 32660179-32660179 on Build GRCh38
rs3204428 Chr.6: 32660192-32660192 on Build GRCh38
rs570286881 Chr.6: 32660100-32660100 on Build GRCh38
rs36205178 Chr.6: 32660161-32660161 on Build GRCh38

View Full Product Details