Assay Design



Gene Transcripts

Gene Symbol
ANO9
Entrez Gene ID
338440
Gene Name
anoctamin 9
Gene Aliases
PIG5, TMEM16J, TP53I5
Chromosome Location
Chr.11: 417930 - 442011 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012302.2 NP_001012302.2 21 - 22 2119 86
XM_011520053.2 XP_011518355.1 21 - 22 2215 86
GenBank mRNA AK026923.1 - 21 - 22 2066 86
AK074088.1 - 14 - 15 4736 86
AK096874.1 - 12 - 13 1407 86
BC128557.1 - 21 - 22 2087 86
BC144415.1 - 20 - 21 2145 86
U33271.1 - 1 - 2 98 85

View Full Product Details