Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47216877-47217445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDB2
Entrez Gene ID
1643
Gene Name
damage specific DNA binding protein 2
Gene Aliases
DDBB, UV-DDB2, XPE
UniGene
Hs.700338
Gene Chromosome Location
Chr.11: 47214942-47239218 on Build GRCh38
Transcripts
NM_000107.2 NM_001300734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
354985 c.293T>C p.L98P Chr.11: 47216886-47216886 on Build GRCh38
7363914 c.304C>T p.R102W Chr.11: 47216897-47216897 on Build GRCh38
4033131 c.382G>A p.A128T Chr.11: 47216975-47216975 on Build GRCh38
6340893 c.433G>T p.D145Y Chr.11: 47217026-47217026 on Build GRCh38
687995 c.292C>G p.L98V Chr.11: 47216885-47216885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529323758 Chr.11: 47217113-47217113 on Build GRCh38
rs4647712 Chr.11: 47217267-47217267 on Build GRCh38
rs55845056 Chr.11: 47217155-47217155 on Build GRCh38
rs201531216 Chr.11: 47217069-47217069 on Build GRCh38
rs539784579 Chr.11: 47217307-47217307 on Build GRCh38
rs78651238 Chr.11: 47216916-47216916 on Build GRCh38
rs780555193 Chr.11: 47216922-47216922 on Build GRCh38
rs147297424 Chr.11: 47217362-47217362 on Build GRCh38
rs781721341 Chr.11: 47217073-47217073 on Build GRCh38
rs886228510 Chr.11: 47217344-47217344 on Build GRCh38
rs1036395849 Chr.11: 47217221-47217221 on Build GRCh38
rs747774067 Chr.11: 47216950-47216950 on Build GRCh38
rs55760739 Chr.11: 47217166-47217166 on Build GRCh38
rs55811602 Chr.11: 47217203-47217203 on Build GRCh38
rs1018189280 Chr.11: 47217311-47217311 on Build GRCh38
rs998067124 Chr.11: 47217335-47217335 on Build GRCh38
rs754091903 Chr.11: 47216906-47216906 on Build GRCh38
rs769144378 Chr.11: 47216946-47216946 on Build GRCh38
rs541629437 Chr.11: 47216911-47216911 on Build GRCh38
rs56367299 Chr.11: 47217165-47217165 on Build GRCh38
rs55642740 Chr.11: 47217202-47217202 on Build GRCh38
rs746180649 Chr.11: 47217378-47217379 on Build GRCh38
rs965708912 Chr.11: 47217325-47217325 on Build GRCh38
rs968226255 Chr.11: 47217419-47217419 on Build GRCh38
rs190052956 Chr.11: 47217092-47217092 on Build GRCh38
rs774225296 Chr.11: 47217096-47217096 on Build GRCh38
rs925732812 Chr.11: 47217358-47217358 on Build GRCh38
rs570718428 Chr.11: 47217286-47217286 on Build GRCh38
rs56167102 Chr.11: 47217172-47217172 on Build GRCh38
rs182352408 Chr.11: 47217144-47217144 on Build GRCh38
rs11537594 Chr.11: 47216912-47216912 on Build GRCh38
rs762943470 Chr.11: 47216963-47216963 on Build GRCh38
rs549726695 Chr.11: 47217057-47217057 on Build GRCh38
rs1024337564 Chr.11: 47217410-47217410 on Build GRCh38
rs145264146 Chr.11: 47217043-47217043 on Build GRCh38
rs1048824403 Chr.11: 47217334-47217334 on Build GRCh38
rs747546269 Chr.11: 47217066-47217066 on Build GRCh38
rs747323146 Chr.11: 47216929-47216929 on Build GRCh38
rs750530703 Chr.11: 47217036-47217036 on Build GRCh38
rs1012514944 Chr.11: 47217388-47217388 on Build GRCh38
rs766359606 Chr.11: 47216969-47216969 on Build GRCh38
rs776380322 Chr.11: 47216947-47216947 on Build GRCh38
rs55749629 Chr.11: 47217204-47217204 on Build GRCh38
rs55884393 Chr.11: 47217130-47217130 on Build GRCh38
rs1049188569 Chr.11: 47217258-47217258 on Build GRCh38
rs326211 Chr.11: 47217114-47217114 on Build GRCh38
rs376867947 Chr.11: 47217091-47217091 on Build GRCh38
rs767324814 Chr.11: 47216972-47216972 on Build GRCh38
rs758898579 Chr.11: 47216914-47216914 on Build GRCh38
rs747801559 Chr.11: 47217078-47217078 on Build GRCh38
rs769399278 Chr.11: 47217084-47217084 on Build GRCh38
rs757474556 Chr.11: 47217168-47217168 on Build GRCh38
rs770431096 Chr.11: 47217095-47217095 on Build GRCh38
rs769321481 Chr.11: 47216958-47216958 on Build GRCh38
rs4647711 Chr.11: 47217112-47217112 on Build GRCh38
rs1004542782 Chr.11: 47217357-47217357 on Build GRCh38
rs921798712 Chr.11: 47217301-47217301 on Build GRCh38
rs759521227 Chr.11: 47217101-47217101 on Build GRCh38
rs760697461 Chr.11: 47217420-47217420 on Build GRCh38
rs139968905 Chr.11: 47216991-47216991 on Build GRCh38
rs533945496 Chr.11: 47217157-47217157 on Build GRCh38
rs944641794 Chr.11: 47217272-47217272 on Build GRCh38
rs896708682 Chr.11: 47217376-47217376 on Build GRCh38
rs992104838 Chr.11: 47217392-47217392 on Build GRCh38
rs772637982 Chr.11: 47216962-47216962 on Build GRCh38
rs56130625 Chr.11: 47217191-47217191 on Build GRCh38
rs56222682 Chr.11: 47217158-47217158 on Build GRCh38
rs994902006 Chr.11: 47217254-47217254 on Build GRCh38
rs533719031 Chr.11: 47217200-47217200 on Build GRCh38
rs200856208 Chr.11: 47216993-47216993 on Build GRCh38
rs765357628 Chr.11: 47217028-47217028 on Build GRCh38
rs757461854 Chr.11: 47217001-47217001 on Build GRCh38
rs326212 Chr.11: 47216971-47216971 on Build GRCh38
rs897834328 Chr.11: 47217223-47217223 on Build GRCh38

View Full Product Details