Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289050.1 | NP_001275979.1 | 5 - 6 | 956 | 94 |
NM_016609.4 | NP_057693.3 | 6 - 7 | 1348 | 94 | |
NM_020372.3 | NP_065105.2 | 5 - 6 | 1081 | 94 | |
XM_005267747.4 | XP_005267804.1 | 6 - 7 | 3424 | 94 | |
XM_017021361.1 | XP_016876850.1 | 6 - 7 | 3421 | 94 | |
XM_017021362.1 | XP_016876851.1 | 6 - 7 | 3419 | 94 | |
GenBank mRNA | BC020565.1 | - | 5 - 6 | 900 | 94 |
BC090870.1 | - | 5 - 6 | 632 | 94 | |
BC111015.1 | - | 4 - 5 | 495 | 94 | |
BX161416.1 | - | 6 - 7 | 1307 | 94 | |
DQ658848.1 | - | 5 - 6 | 726 | 94 |