Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014696.3 | NP_055511.2 | 2 - 3 | 127 | 140 |
XM_005277666.3 | XP_005277723.1 | 3 - 4 | 403 | 140 | |
XM_011540400.2 | XP_011538702.1 | 2 - 3 | 248 | 140 | |
GenBank mRNA | BC011672.2 | - | 2 - 3 | 127 | 140 |